Merck
  • Development of alginate-reinforced chitosan nanoparticles utilizing W/O nanoemulsification/internal crosslinking technique for transdermal delivery of rabeprazole.
Development of alginate-reinforced chitosan nanoparticles utilizing W/O nanoemulsification/internal crosslinking technique for transdermal delivery of rabeprazole.
Life sciences (2014-07-06)
Tarek A Ahmed, Khalid M El-Say
ABSTRACT
First; to develop rabeprazole (RP)-alginate core coated chitosan nanoparticles (NP) utilizing water in oil (W/O) nanoemulsion technique. Second; formulation of transdermal patches loaded RP-NP that avoid drug peroral acid sensitivity and first pass effect. The influence of six factors on RP-NP formulation was investigated using Plackett-Burman (PB) design. The studied factors were considered for their effect on particle size (Y1) and loading efficiency (Y2). Formulation optimum desirability was identified; a proposed formulation was prepared and characterized. In vitro permeation of the prepared NP compared with RP was studied. Transdermal patches loaded drug or RP-NP were prepared and characterized. Patches ex vivo permeation through rat skin was studied, and kinetic analysis and permeation mechanism were investigated. Chitosan, oil phase and surfactant to oil ratios had significant effects on Y1, while Y2 was significantly affected by the same variables affecting Y1 and span80-tween80 ratio. Scanning electron microscope imaging illustrated sphericity of the NP. The optimized RP-NP exhibited sustained release pattern. The prepared patches showed a minimal patch to patch variable. Patches loaded RP-NP exhibited substantial skin permeability and controlled drug release, and were in favor of Fickian diffusion. Transdermal patches loaded RP-NP is effective drug delivery and alternative to drug peroral route.
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Calcium chloride
Sigma-Aldrich
Calcium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
1,2-Propanediol, meets analytical specification of Ph. Eur., BP, USP, ≥99.5%
Supelco
1,2-Propanediol, analytical standard
Sigma-Aldrich
1,2-Propanediol, puriss. p.a., ACS reagent, ≥99.5% (GC)
Sigma-Aldrich
1,2-Propanediol, ReagentPlus®, 99%
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
1,2-Propanediol, ACS reagent, ≥99.5%
Sigma-Aldrich
Hexane, puriss. p.a., ACS reagent, ≥99.0% (GC)
Sigma-Aldrich
1,2-Propanediol, tested according to Ph. Eur.
Supelco
Hexane, analytical standard
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium alginate
Supelco
Calcium ion solution for ISE, 0.1 M Ca, for ion-selective electrodes, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Calcium chloride, Vetec, reagent grade, 96%
Sigma-Aldrich
Hexane, ReagentPlus®, ≥99%
Sigma-Aldrich
Hexane, Laboratory Reagent, ≥95%
Sigma-Aldrich
Hexane, suitable for HPLC, ≥95%
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
Hexane, suitable for HPLC, ≥97.0% (GC)
Sigma-Aldrich
Hexane, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99% (GC)
Sigma-Aldrich
Chitosan from shrimp shells, low-viscous
Sigma-Aldrich
Calcium chloride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Alginic acid from brown algae, powder
Supelco
Propylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Rabeprazole sodium, ≥98% (HPLC)
USP
Propylene glycol, United States Pharmacopeia (USP) Reference Standard