Merck
  • Separation of digesta fractions complicates estimation of ileal digestibility using marker methods with Cr2O3 and cobalt-ethylenediamine tetraacetic acid in broiler chickens.
Separation of digesta fractions complicates estimation of ileal digestibility using marker methods with Cr2O3 and cobalt-ethylenediamine tetraacetic acid in broiler chickens.
Poultry science (2014-06-17)
S de Vries, R P Kwakkel, A M Pustjens, M A Kabel, W H Hendriks, W J J Gerrits
ABSTRACT
Marker methodologies to measure ileal and total tract digestibilities of diets varying in content and degradability of dietary fiber in broiler chickens were evaluated. Chromium sesquioxide (Cr2O3) and cobalt-ethylenediamine tetraacetic acid (Co-EDTA) were used as markers of solid and soluble fractions, respectively, and compared with digestibility values obtained with the total collection method. Groups of broilers (n = 17, 11 broilers/group) were assigned to a low-fiber diet or 1 of 2 high-fiber diets, the latter 2 containing 35% rapeseed meal (RSM). Pectolytic enzymes were added to one RSM diet to improve degradability of the fiber fraction. Excreta were quantitatively collected for 96 h, and contents from ileum and ceca were collected at slaughter at 29, 30, or 31 d of age. Chromium recovery in excreta ranged between 86 and 95%, whereas cobalt recovery was considerably lower (66 to 70%). Chromium:cobalt ratio was higher in ileal digesta than in feed. Hardly any chromium was found in the ceca, indicating that separation of the marker and specific digesta fractions occurs. Estimates of apparent total tract digestibility (ATTD) were lower when calculated using the marker method compared with the total collection method, particularly in high-fiber diets. Using Cr2O3 as a marker, differences were relatively small and effects due to enzyme addition were generally similar. Using Co-EDTA as a marker, ATTD of all components were lower compared with values obtained using the collection method (3 to 45 percentage units, P < 0.001), likely related to the low Co recovery. When estimating apparent ileal digestibility (AID), separation of marker and digesta resulted in unrealistically high estimates for the digestibility of nonglucose polysaccharides (54 to 66%), exceeding ATTD values by 16 to 42 percentage units. Moreover, the effect of pectolytic enzyme addition on the AID of nonglucose polysaccharides was in the opposite direction when compared with total collection. The data illustrate that fractionation of digesta, particularly in high-fiber diets, complicates accurate AID measurements in broilers, regardless of the choice of markers used.
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, BioUltra, for molecular biology, ≥99.0% (T)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, meets analytical specification of Ph. Eur., BP, USP, FCC, 99.0-101.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, ≥97.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, Sigma Grade, suitable for plant cell culture, 98.5-101.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, reagent grade, 98.5-101.5% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, 98.5-101.5%, BioUltra
Sigma-Aldrich
Ethylenediaminetetraacetic acid calcium disodium salt hydrate
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, ACS reagent, 99.0-101.0%
USP
Edetate disodium, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Chromium(III) oxide, nanopowder, <100 nm particle size (TEM), 98% trace metals basis
Supelco
Ethylenediaminetetraacetic acid disodium salt dihydrate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, suitable for electrophoresis, for molecular biology, 99.0-101.0% (titration)
Disodium edetate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Chromium(III) oxide, powder, 99.9% trace metals basis
Sigma-Aldrich
Chromium(III) oxide, powder, ≥98%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, 2%, solution
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sodium calcium edetate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ethylenediaminetetraacetic acid, Vetec, reagent grade, 98%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, for molecular biology, 0.5 M in H2O, DNase, RNase, NICKase and protease, none detected
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)