Merck
  • Induction of hepatic apolipoprotein A-I gene expression by the isoflavones quercetin and isoquercetrin.
Induction of hepatic apolipoprotein A-I gene expression by the isoflavones quercetin and isoquercetrin.
Life sciences (2014-06-26)
Michael J Haas, Luisa M Onstead-Haas, Anna Szafran-Swietlik, Hagop Kojanian, Tim Davis, Paul Armstrong, Norman C W Wong, Arshag D Mooradian
ABSTRACT
Phytochemicals such as flavonoids, vitamins, and polyphenols have been shown to have beneficial effects in metabolic disease. To determine if select flavonoids regulate hepatic apolipoprotein A-I (apo A-I) and high-density lipoprotein (HDL) synthesis, we examined the effects of quercetin, isoquercetin, and myrescetin on apo A-I gene expression in HepG2 (hepatocytes) and Caco-2 (intestinal) cells. Apo A-I gene expression was measured by Western blotting, quantitative reverse-transcription polymerase chain reaction, and transient transfection. Estrogen receptor α (ESR1) and estrogen receptor β expression were measured by Western blotting, and ESR1 expression was inhibited using ESR1-specific short inhibitory RNA (siRNA). Quercetin and isoquercetin, but not myrecetin, induced apo A-I protein and mRNA synthesis, and induced apo A-I promoter activity. Induction by quercetin required an estrogen-responsive region of the apo A-I promoter. Addition of estrogen receptor blocker ICI-182780 to quercetin-treated cells inhibited the effects of quercetin on apo A-I gene expression. Down-regulation of ESR1 with ESR1 siRNA had no effect on basal apo A-I gene expression; however it prevented quercetin-mediated induction of apo A-I gene expression. We conclude that quercetin induces apo A-I gene expression at least in part through induction of ESR1 and may be useful in treating hypoalphalipoproteinemia.
Sigma-Aldrich
β-Estradiol, analytical standard
Sigma-Aldrich
β-Estradiol, BioReagent, powder, suitable for cell culture
Sigma-Aldrich
β-Estradiol, powder, γ-irradiated, suitable for cell culture
Sigma-Aldrich
Quercetin, ≥95% (HPLC), solid
Sigma-Aldrich
Fulvestrant, >98% (HPLC)
Fulvestrant for system suitability, European Pharmacopoeia (EP) Reference Standard
Fulvestrant, European Pharmacopoeia (EP) Reference Standard
Isoquercitroside, European Pharmacopoeia (EP) Reference Standard
Supelco
Myricetin, analytical standard
Sigma-Aldrich
Myricetin, ≥96.0% (HPLC)
Sigma-Aldrich
Estradiol, meets USP testing specifications
Sigma-Aldrich
Myricetin, ≥96.0%, crystalline
Sigma-Aldrich
β-Estradiol, ≥98%
USP
Estradiol, United States Pharmacopeia (USP) Reference Standard
Estradiol hemihydrate, European Pharmacopoeia (EP) Reference Standard
Supelco
Estradiol, Pharmaceutical Secondary Standard; Certified Reference Material