Merck
  • Gastric emptying is involved in Lactobacillus colonisation in mouse stomach.
Gastric emptying is involved in Lactobacillus colonisation in mouse stomach.
The British journal of nutrition (2014-06-17)
Yuraporn Sahasakul, Naoki Takemura, Kei Sonoyama
ABSTRACT
Lactobacilli are indigenous microbes of the stomach of rodents, with much lower numbers being present in mice fed a purified diet than in those fed a non-purified diet. We postulated that gastric emptying (GE) is responsible for the different colonisation levels of lactobacilli and tested this hypothesis in the present study. BALB/cCr Slc mice were fed either a non-purified diet or a purified diet for 2 weeks. The number of gastric tissue-associated lactobacilli was lower in mice fed the purified diet than in those fed the non-purified diet. GE, estimated by measuring the food recovered from the stomach, was higher in mice fed the purified diet than in those fed the non-purified diet and correlated negatively with the number of lactobacilli. Mice fed the non-purified diet exhibited lower GE rates even when lactobacilli were eliminated by ampicillin administration through the drinking-water, suggesting that GE is the cause but not the consequence of different Lactobacillus colonisation levels. The plasma concentrations of acylated ghrelin, a gastric hormone that promotes GE, were higher in mice fed the purified diet than in those fed the non-purified diet. There was a negative correlation between GE and the number of lactobacilli in mice fed the non-purified diet, the purified diet, and the purified diet supplemented with sugarbeet fibre (200 g/kg diet) or carboxymethyl cellulose (40 g/kg diet). We propose that a higher GE rate contributes, at least in part, to lower gastric colonisation levels of lactobacilli in mice fed a purified diet.
Product Description
Supelco
Ampicillin trihydrate, VETRANAL®, analytical standard
Sigma-Aldrich
Ampicillin, meets USP testing specifications
Sigma-Aldrich
Ampicillin trihydrate, 900-1050 μg/mg anhydrous basis (HPLC)
Ampicillin trihydrate, European Pharmacopoeia (EP) Reference Standard
Supelco
Ampicillin Trihydrate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ampicillin sodium salt, BioXtra, suitable for cell culture
Sigma-Aldrich
Ampicillin sodium salt
Sigma-Aldrich
Ampicillin, anhydrous, 96.0-102.0% (anhydrous basis)
Sigma-Aldrich
Ampicillin sodium salt, powder or crystals, BioReagent, suitable for cell culture
Ampicillin, anhydrous, European Pharmacopoeia (EP) Reference Standard
USP
Ampicillin, United States Pharmacopeia (USP) Reference Standard
Supelco
Ampicillin Sodium, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ampicillin, analytical standard