Merck
  • QM/MM study of the reaction mechanism of the carboxyl transferase domain of pyruvate carboxylase from Staphylococcus aureus.
QM/MM study of the reaction mechanism of the carboxyl transferase domain of pyruvate carboxylase from Staphylococcus aureus.
Biochemistry (2014-06-26)
Xiang Sheng, Yongjun Liu
ABSTRACT
Pyruvate carboxylase (PC) catalyzes the carboxylation of pyruvate to produce oxaloacetate. Its activity is directly related to insulin release and thus PC has recently attracted great interest as a potential target for diabetes treatment. In this article, the catalytic mechanism of the carboxyl transferase domain of PC from Staphylococcus aureus was investigated by using a combined quantum-mechanical/molecular-mechanical approach. Our calculation results indicate that the catalytic reaction starts from the decarboxylation of carboxybiotin to generate an enol-BTI intermediate, followed by two consecutive proton-transfer processes (from T908 to enol-BTI and from PYR to T908). During the catalytic reaction, the main-chain amide of T908 plays a key role in catching CO2 and preventing its diffusion from the active center. A triad of residues, R571, Q575, and K741, contributes both to substrate binding and enol-pyruvate stabilization. The oxyanion hole, consisting of the side-chain hydroxyl of S911 and the side-chain amino of Q870, plays an important role in stabilizing the hydroxyl anion of BTI. The coordination of the metal cation by pyruvate is a second sphere, rather than an inner sphere, interaction, and the metal cation stabilizes the species through the medium of residue K741. The decarboxylation of carboxybiotin corresponds to the highest free energy barrier of 21.7 kcal/mol. Our results may provide useful information for both the regulation of enzyme activity and the development of related biocatalytic applications.
Sigma-Aldrich
Zinc, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.9%, granular
Sigma-Aldrich
Pyruvate Carboxylase from bovine liver, buffered aqueous glycerol solution, 5-25 units/mg protein (BCA)
Sigma-Aldrich
Zinc, foil, thickness 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Zinc, pieces, 2-14 mesh, 99.9% trace metals basis
Sigma-Aldrich
Zinc, dust, <10 μm, ≥98%
Sigma-Aldrich
Manganese, powder, ≥99.9% trace metals basis
Sigma-Aldrich
Zinc preparation, 5 g/dL Zn+2 in THF, highly reactive Rieke®metal
Sigma-Aldrich
Manganese, chips, thickness <2.0 mm, 99%
Sigma-Aldrich
Zinc, foil, thickness 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Zinc, shot, 5 mm, 99.999% trace metals basis
Sigma-Aldrich
Zinc, granular, 20-30 mesh, ACS reagent, ≥99.8%
Sigma-Aldrich
Zinc, sticks, diam. 7-10 mm, 99.97% trace metals basis
Sigma-Aldrich
Manganese, powder, −325 mesh, ≥99% trace metals basis
Sigma-Aldrich
Zinc, nanopowder, 40-60 nm avg. part. size, ≥99% trace metals basis
Sigma-Aldrich
Zinc, shot, <12 mm, 99.99% trace metals basis
Zinc, foil, 6mm disks, thickness 0.125mm, as rolled, 99.99+%
Zinc, foil, light tested, 25x25mm, thickness 0.025mm, as rolled, 99.99+%
Zinc, foil, 6mm disks, thickness 1.0mm, as rolled, 99.95+%
Zinc, foil, 8mm disks, thickness 0.20mm, as rolled, 99.95+%
Zinc, foil, 6mm disks, thickness 0.015mm, 99.9%
Zinc, foil, not light tested, 150x150mm, thickness 0.025mm, as rolled, 99.95+%
Zinc, foil, light tested, 150x150mm, thickness 0.025mm, as rolled, 99.95+%
Zinc, foil, not light tested, 100x100mm, thickness 0.025mm, as rolled, 99.99+%
Zinc, foil, 5m coil, thickness 0.25mm, as rolled, 99.95+%
Zinc, foil, light tested, 50x50mm, thickness 0.05mm, as rolled, 99.95+%
Zinc, foil, 8mm disks, thickness 0.35mm, as rolled, 99.95+%
Zinc, foil, 6mm disks, thickness 1.0mm, as rolled, 99.99+%
Zinc, foil, 6mm disks, thickness 0.01mm, 99.9%
Zinc, foil, light tested, 150x150mm, thickness 0.05mm, as rolled, 99.95+%
Zinc, foil, 6mm disks, thickness 0.175mm, as rolled, 99.95+%