Merck
  • Carvedilol has stronger anti-inflammation and anti-virus effects than metoprolol in murine model with coxsackievirus B3-induced viral myocarditis.
Carvedilol has stronger anti-inflammation and anti-virus effects than metoprolol in murine model with coxsackievirus B3-induced viral myocarditis.
Gene (2014-06-07)
Dan Wang, Yiming Chen, Jianbin Jiang, Aihua Zhou, Lulu Pan, Qi Chen, Yan Qian, Maoping Chu, Chao Chen
ABSTRACT
This study aims to compare the effects of carvedilol and metoprolol in alleviating viral myocarditis (VMC) induced by coxsackievirus B3 (CVB3) in mice. A total of 116 Balb/c mice were included in this study. Ninety-six mice were inoculated intraperitoneally with CVB3 to induce VMC. The CVB3 inoculated mice were evenly divided into myocarditis group (n=32), carvedilol group (n=32) and metoprolol group (n=32). Twenty mice (control group) were inoculated intraperitoneally with normal saline. Hematoxylin and eosin staining and histopathologic scoring were used to investigate the effects of carvedilol and metoprolol on myocardial histopathologic changes on days 3 and 5. In addition, serum cTn-I levels, cytokine levels and virus titers were determined using chemiluminescence immunoassay, enzyme-linked immunosorbent assay and plaque assay, respectively, on days 3 and 5. Finally, the levels of phosphorylated p38MAPK were studied using immunohistochemical staining and Western blotting on day 5. Carvedilol had a stronger effect than metoprolol in reducing the pathological scores of VMC induced by CVB3. Both carvedilol and metoprolol reduced the levels of cTn-I, but the effect of carvedilol was stronger. Carvedilol and metoprolol decreased the levels of myocardial pro-inflammatory cytokines and increased the expression of anti-inflammatory cytokine, with the effects of carvedilol being stronger than those of metoprolol. Carvedilol had a stronger effect in reducing myocardial virus concentration compared with metoprolol. Carvedilol was stronger than metoprolol in decreasing the levels of myocardial phosphorylated p38MAPK. In conclusion, carvedilol was more potent than metoprolol in ameliorating myocardial lesions in VMC, probably due to its stronger modulation of the balance between pro- and anti-inflammatory cytokines by inhibiting the activation of p38MAPK pathway through β1- and β2-adrenoreceptors.
Product Description
Supelco
Metoprolol Tartrate, Pharmaceutical Secondary Standard; Certified Reference Material
Metoprolol tartrate, European Pharmacopoeia (EP) Reference Standard
USP
Metoprolol tartrate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Carvedilol, ≥98% (HPLC), solid
Sigma-Aldrich
(±)-Metoprolol (+)-tartrate salt, ≥98% (titration), powder
Carvedilol for system suitability, European Pharmacopoeia (EP) Reference Standard
USP
Carvedilol, United States Pharmacopeia (USP) Reference Standard
Supelco
Carvedilol, Pharmaceutical Secondary Standard; Certified Reference Material
Carvedilol, European Pharmacopoeia (EP) Reference Standard