Merck
Percutaneous bone lesion ablation.
La Radiologia medica (2014-06-05)
Dimitrios K Filippiadis, Sean Tutton, Alexis Kelekis
ABSTRACT
Benign tumors and metastatic bone lesions can be treated by ablation techniques performed either alone or in combination with other percutaneous techniques. Ablation techniques include ethanol or acetic acid injection and thermal ablation by means of energy deposition [including laser, radiofrequency, microwave, cryoablation, radiofrequency ionization and magnetic resonance (MR)-guided high-intensity focused ultrasound (HIFU)]. Goal definition of the therapy is crucial: ablation techniques can be proposed as curative treatments in benign bone tumors or oligometastatic disease (<3 lesions). Alternatively, these techniques can be proposed as palliative treatments aiming at reduction of pain, local control of the disease and tumor decompression. Depending on the lesion's location ablation can be combined with cementation with or without further metallic augmentation; local tumor control can be enhanced by combining ablation with transarterial bland embolization or chemoembolization. Thermal ablation of bone and soft tissues is characterized by high success and relatively low rates of potential complications, mainly iatrogenic thermal damage of surrounding sensitive structures. Successful thermal ablation requires a sufficient ablation volume and thermal protection of the surrounding vulnerable structures. This article will describe the general principles governing ablation and the mechanism of action for each technique and in addition will review the literature about safety and effectiveness of percutaneous imaging-guided ablation for benign and malignant (primary and metastatic) lesions.
Product Description
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Reagent Alcohol, reagent grade
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.005% water
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.003% water
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, glacial, puriss., 99-100%
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Reagent Alcohol, denatured, suitable for HPLC
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 2% 2-butanone, A15 MEK1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
USP
Glacial acetic acid, United States Pharmacopeia (USP) Reference Standard
Millipore
Bifido Selective Supplement B, suitable for microbiology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof
Sigma-Aldrich
Ethanol, ACS reagent, prima fine spirit, without additive, F15 o1
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%