Merck
  • Feeling unreal: a functional imaging study in patients with Kleine-Levin syndrome.
Feeling unreal: a functional imaging study in patients with Kleine-Levin syndrome.
Brain : a journal of neurology (2014-05-03)
Aurelie Kas, Sophie Lavault, Marie-Odile Habert, Isabelle Arnulf
ABSTRACT
Kleine-Levin syndrome is characterized by relapsing-remitting episodes of severe hypersomnia, cognitive impairment, apathy, derealization and behavioural disturbances. Between episodes, patients have normal sleep, mood and behaviour. Functional imaging studies performed in small series of patients with Kleine-Levin syndrome with visual or semi-quantitative, uncontrolled analysis yielded equivocal brain changes. Using whole brain voxel-based group analysis, we compared brain perfusion scintigraphy during and between episodes in consecutive patients with Kleine-Levin syndrome versus healthy control subjects and correlated perfusion changes with disease severity and symptoms, focusing on less studied but disabling symptoms, such as apathy and derealization. During asymptomatic periods, 41 patients (mean age of 22.3 ± 8.1 years, 56.1% male) and 15 age- and sex-matched healthy control subjects underwent single-photon emission computed tomography scanning with technetium-99m ethyl cysteinate dimer. Eleven patients repeated the test during a symptomatic period. Compared with controls, patients during asymptomatic periods had persistent hypoperfusion in the hypothalamus, the thalamus (mainly the right posterior part), the caudate nucleus, and cortical associative areas, including the anterior cingulate, (Brodmann area 25), the orbito-frontal (Brodmann area 11) and the right superior temporal cortices (Brodmann area 22), extending to the insula (P < 0.001 in all area). Two additional hypoperfused areas emerged during symptomatic periods (P < 0.001), located in the right dorsomedial prefrontal cortex (Brodmann area 8) and the right parieto-temporal junction (Brodmann areas 22 and 39). These two areas were more affected between episodes, when the mean episode duration was longer (r = -0.53; P < 0.001). The score for the Depersonalization/Derealization Inventory during symptomatic periods strongly correlated with the hypoperfusion of the right (r = -0.74, P < 0.001) and left (r = -0.59, P < 0.005) parieto-temporal junctions. No hyperperfusion was found. Because the parieto-temporal junction (including the angular gyrus) is involved in cross-modal association between somatosensory (body knowledge), auditory and visual information, the robust hypoperfusions and correlations observed in this area may underlie the striking derealization reported by patients during episodes. Defects in the dorsomedial prefrontal cortex may cause apathy. Persistent hypoperfusion in the diencephalic and associative cortical area during asymptomatic periods is a marker of the disease, suggestive of a scenario wherein patients compensate for these deficient circuitries.
Product Description
Supelco
L-Cysteine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Cysteine, Wacker Chemie AG, ≥98.0%
SAFC
L-Cysteine
Sigma-Aldrich
L-Cysteine, ≥97%, FG
Sigma-Aldrich
L-Cysteine, Vetec, reagent grade, 97%
Sigma-Aldrich
L-Cysteine, 97%
Sigma-Aldrich
L-Cysteine, BioUltra, ≥98.5% (RT)
Sigma-Aldrich
L-Cysteine, from non-animal source, BioReagent, suitable for cell culture, ≥98%