Merck
  • Study of photocatalytic degradation of tributyltin, dibutylin and monobutyltin in water and marine sediments.
Study of photocatalytic degradation of tributyltin, dibutylin and monobutyltin in water and marine sediments.
Chemosphere (2014-03-13)
Stephan Brosillon, Chrystelle Bancon-Montigny, Julie Mendret
ABSTRACT
This study reports on the first assessment of the treatment of sediments contaminated by organotin compounds using heterogeneous photocatalysis. Photocatalysis of organotins in water was carried out under realistic concentration conditions (μgL(-1)). Degradation compounds were analyzed by GC-ICP-MS; a quasi-complete degradation of tributyltin (TBT) in water (99.8%) was achieved after 30min of photocatalytic treatment. The degradation by photolysis was about (10%) in the same conditions. For the first time decontamination of highly polluted marine sediments (certified reference material and harbor sediments) by photocatalysis proves that the use of UV and the production of hydroxyl radicals are an efficient way to treat organotins adsorbed onto marine sediment despite the complexity of the matrix. In sediment, TBT degradation yield ranged from 32% to 37% after only 2h of irradiation (TiO2-UV) and the by-products: dibutyltin (DBT) and monobutyltin (MBT) were degraded very rapidly in comparison with TBT. It was shown that during photocatalysis of organotins in sediments, the hydroxyl radical attack and photolysis are the two ways for the degradation of adsorbed TBT.
Sigma-Aldrich
Titanium(IV) oxide, rutile, ≥99.98% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, 99.995% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, powder, <5 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, mixture of rutile and anatase, nanoparticles, <150 nm particle size (volume distribution, DLS), dispersion, 40 wt. % in H2O, 99.5% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, nanopowder, <100 nm particle size, 99.5% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, <001>, (single crystal substrate), ≥99.9% trace metals basis, L × W × thickness 10 mm × 10 mm × 0.5 mm
Sigma-Aldrich
Titanium(IV) oxide, mixture of rutile and anatase, nanopowder, <100 nm particle size (BET), 99.5% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, nanowires, diam. × L ~100 nm × 10 μm
Sigma-Aldrich
Titanium(IV) oxide, anatase, nanopowder, <25 nm particle size, 99.7% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, nanopowder, 21 nm primary particle size (TEM), ≥99.5% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, nanowires, diam. × L ~10 nm × 10 μm
Sigma-Aldrich
Titanium(IV) oxide, contains 1% Mn as dopant, nanopowder, <100 nm particle size (BET), ≥97%
Sigma-Aldrich
Titanium(IV) oxide, anatase, powder, 99.8% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, anatase, powder, −325 mesh, ≥99% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, puriss., meets analytical specification of Ph. Eur., BP, USP, 99-100.5%
Sigma-Aldrich
Titanium(IV) oxide, ReagentPlus®, ≥99%