Merck
  • Persistent infectious diseases say - IDO. Role of indoleamine-2,3-dioxygenase in disease pathogenesis and implications for therapy.
Persistent infectious diseases say - IDO. Role of indoleamine-2,3-dioxygenase in disease pathogenesis and implications for therapy.
Critical reviews in microbiology (2012-11-24)
Heidi Barth, Sukanya Raghuraman
ABSTRACT
Indoleamine-2,3-dioxygenase (IDO) is an enzyme that catabolises tryptophan - an essential amino acid critical for T cell proliferation. Initially recognized as a first line of host defense against infectious pathogens, IDO has been subsequently identified as an important immune-regulator inhibiting T-cell responses and promoting immune tolerance. Research over the past few years has demonstrated a crucial role for IDO in the pathogenesis of persistent infections that place an enormous burden on public health. In this review, we summarize current knowledge about IDO's role in causing pathogen persistence and progression to clinical disease. We conclude with a perspective on the potential benefits and risks of therapeutic IDO manipulation.
Product Description
Supelco
Tryptophan, Pharmaceutical Secondary Standard; Certified Reference Material
Tryptophan, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Tryptophan, BioUltra, ≥99.5% (NT)
Supelco
L-Tryptophan, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Tryptophan, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Tryptophan, Vetec, reagent grade, ≥98%
Sigma-Aldrich
L-Tryptophan, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%