Merck
  • Early empiric antibiotic use in preterm infants is associated with lower bacterial diversity and higher relative abundance of Enterobacter.
Early empiric antibiotic use in preterm infants is associated with lower bacterial diversity and higher relative abundance of Enterobacter.
The Journal of pediatrics (2014-02-18)
Corryn Greenwood, Ardythe L Morrow, Anne J Lagomarcino, Mekibib Altaye, Diana H Taft, Zhuoteng Yu, David S Newburg, Doyle V Ward, Kurt R Schibler
ABSTRACT
To determine the impact of empiric ampicillin and gentamicin use in the first week of life on microbial colonization and diversity in preterm infants. The 16s ribosomal DNA community profiling was used to compare the microbiota of 74 infants born ≤32 weeks gestational age by degree of antibiotic use in the first week of life. The degree of antibiotic use was classified as 0 days, 1-4 days, and 5-7 days of antibiotic administration. All of the antibiotic use was empiric, defined as treatment based solely on clinical suspicion of infection without a positive culture result. Infants who received 5-7 days of empiric antimicrobial agents in the first week had increased relative abundance of Enterobacter (P = .016) and lower bacterial diversity in the second and third weeks of life. Infants receiving early antibiotics also experienced more cases of necrotizing enterocolitis, sepsis, or death than those not exposed to antibiotics. Early empiric antibiotics have sustained effects on the intestinal microbiota of preterm infants. Intestinal dysbiosis in this population has been found to be associated with elevated risk of necrotizing enterocolitis, sepsis, or death.
Product Description
Supelco
Chloroform, analytical standard
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Supelco
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Ampicillin, meets USP testing specifications
Sigma-Aldrich
Ampicillin trihydrate, 900-1050 μg/mg anhydrous basis (HPLC)
Supelco
Ampicillin trihydrate, VETRANAL®, analytical standard
Supelco
Ampicillin Trihydrate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Ampicillin trihydrate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, puriss. p.a., reag. ISO, reag. Ph. Eur., 99.0-99.4% (GC)
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, amylene stabilized
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Ampicillin, anhydrous, 96.0-102.0% (anhydrous basis)
Sigma-Aldrich
Ampicillin sodium salt, powder or crystals, BioReagent, suitable for cell culture
Sigma-Aldrich
Ampicillin sodium salt
Supelco
Residual Solvent - Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ampicillin sodium salt, BioXtra, suitable for cell culture