Merck
  • A decrease of the butyrate-producing species Roseburia hominis and Faecalibacterium prausnitzii defines dysbiosis in patients with ulcerative colitis.
A decrease of the butyrate-producing species Roseburia hominis and Faecalibacterium prausnitzii defines dysbiosis in patients with ulcerative colitis.
Gut (2013-09-12)
Kathleen Machiels, Marie Joossens, João Sabino, Vicky De Preter, Ingrid Arijs, Venessa Eeckhaut, Vera Ballet, Karolien Claes, Filip Van Immerseel, Kristin Verbeke, Marc Ferrante, Jan Verhaegen, Paul Rutgeerts, Séverine Vermeire
ABSTRACT
Bacteria play an important role in the onset and perpetuation of intestinal inflammation in inflammatory bowel disease (IBD). Unlike in Crohn's disease (CD), in which dysbiosis has been better characterised, in ulcerative colitis (UC), only small cohorts have been studied and showed conflicting data. Therefore, we evaluated in a large cohort if the microbial signature described in CD is also present in UC, and if we could characterise predominant dysbiosis in UC. To assess the functional impact of dysbiosis, we quantified the bacterial metabolites. The predominant microbiota from 127 UC patients and 87 age and sex-matched controls was analysed using denaturing gradient gel electrophoresis (DGGE) analysis. Differences were quantitatively validated using real-time PCR. Metabolites were quantified using gas chromatography-mass spectrometry. Based on DGGE analysis, the microbial signature previously described in CD was not present in UC. Real-time PCR analysis revealed a lower abundance of Roseburia hominis (p<0.0001) and Faecalibacterium prausnitzii (p<0.0001) in UC patients compared to controls. Both species showed an inverse correlation with disease activity. Short-chain fatty acids (SCFA) were reduced in UC patients (p=0.014), but no direct correlation between SCFA and the identified bacteria was found. The composition of the fecal microbiota of UC patients differs from that of healthy individuals: we found a reduction in R hominis and F prausnitzii, both well-known butyrate-producing bacteria of the Firmicutes phylum. These results underscore the importance of dysbiosis in IBD but suggest that different bacterial species contribute to the pathogenesis of UC and CD.
Sigma-Aldrich
Sodium sulfate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Supelco
Sulfuric acid, for the determination of nitrogen, ≥97.0%
Sigma-Aldrich
Sodium sulfate, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Sodium sulfate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Sodium sulfate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sulfuric acid, puriss. p.a., for determination of Hg, ACS reagent, reag. ISO, reag. Ph. Eur., 95.0-97.0%
Sigma-Aldrich
Sulfuric acid, puriss., meets analytical specification of Ph. Eur., BP, 95-97%
Sigma-Aldrich
Sodium sulfate, puriss., meets analytical specification of Ph. Eur., BP, USP, anhydrous, 99.0-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, powder
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, granular
Sigma-Aldrich
Sulfuric acid, ACS reagent, 95.0-98.0%
Sigma-Aldrich
Sodium sulfate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Sulfuric acid solution, puriss. p.a., ≥25% (T)
Sigma-Aldrich
Propionic acid, ≥99.5%
Supelco
Butyric acid, analytical standard
Sigma-Aldrich
Butyric acid, ≥99%
Sigma-Aldrich
Butyric acid, natural, ≥99%, FCC, FG
Sigma-Aldrich
Propionic acid, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Propionic acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Propionic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Propionic acid, natural, 99%, FG
Sigma-Aldrich
Butyric acid, ≥99%, FG
Sigma-Aldrich
Propionic acid, BioReagent, suitable for insect cell culture, ~99%
Supelco
Propionic acid, analytical standard
Supelco
Sulfuric acid concentrate, 0.1 M H2SO4 in water (0.2N), eluent concentrate for IC