Merck
Get up to 22% off for Pi Day until 3/26.Save Now
6-[4-Chloro-2-(trifluoro-meth-yl)phen-yl]-3-fluoro-2-methyl-pyridine.
Acta crystallographica. Section E, Structure reports online (2013-03-12)
S Sreenivasa, K E Manojkumar, P A Suchetan, N R Mohan, B S Palakshamurthy, T Srinivasan, D Velmurgan
ABSTRACT
In the title compound, C13H8ClF4N, the dihedral angle between the benzene and pyridine rings is 59.8 (3)°. In the crystal, mol-ecules are stacked in columns along the b axis through weak C-H⋯π inter-actions.