Merck
  • Construction of L-lysine-, L-threonine-, and L-isoleucine-overproducing strains of Corynebacterium glutamicum.
Construction of L-lysine-, L-threonine-, and L-isoleucine-overproducing strains of Corynebacterium glutamicum.
Annals of the New York Academy of Sciences (1996-05-15)
H Sahm, L Eggeling, B Eikmanns, R Krämer
ABSTRACT
The gram-negative bacterium Corynebacterium glutamicum is used for the industrial production of amino acids, for example, of L-glutamate and L-lysine. By cloning and expressing the various genes of the L-lysine pathway in C. glutamicum, we would demonstrate that an increase of the flux of L-aspartate semialdehyde to L-lysine could be obtained in strains with increased dihydrodipicolinate synthase activity. Recently we detected that in C. glutamicum two pathways exist for synthesis of D,L-diaminopimelate and L-lysine. Mutants defective in one pathway are still able to synthesize enough L-lysine for growth, but the L-lysine secretion is reduced to 50 to 70%. Using NMR spectroscopy, we could calculate how much of the L-lysine secreted into the medium is synthesized via either one or the other pathway. Amplification of the feedback inhibition insensitive homoserine dehydrogenase and homoserine kinase in a high L-lysine-overproducing strain enabled channeling of the carbon flow from the intermediate aspartate semialdehyde towards homoserine, resulting in a high accumulation of L-threonine. For a further flux from L-threonine to L-isoleucine, the allosteric control of threonine dehydratase was eliminated.
Product Description
Supelco
L-Threonine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
SAFC
L-Threonine
Sigma-Aldrich
L-Lysine, crystallized, ≥98.0% (NT)
Sigma-Aldrich
L-Threonine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
L-Lysine, ≥98% (TLC)
Sigma-Aldrich
L-Threonine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Threonine, BioXtra, ≥99.5% (NT)
Isoleucine, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Lysine, analytical standard
L-Threonine, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Threonine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Isoleucine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Isoleucine, 99%, FCC, FG
SAFC
L-Isoleucine
Sigma-Aldrich
L-Isoleucine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Isoleucine, reagent grade, ≥98% (HPLC)
Supelco
L-Isoleucine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Isoleucine, BioUltra, ≥99.5% (NT)