Merck
  • Chemical and microbiologic characteristics and toxicity of povidone-iodine solutions.
Chemical and microbiologic characteristics and toxicity of povidone-iodine solutions.
American journal of surgery (1986-03-01)
J L Zamora
ABSTRACT
The chemical, microbiologic, and toxic characteristics of povidone-iodine solution, a commonly used antiseptic agent, are addressed in a collective review of relevant works published from 1956 to the present. Interactions of this agent with the thyroid gland and the kidneys and its toxicity at the cellular and systemic level are discussed.
Sigma-Aldrich
Kollidon® 25
Supelco
Poly(vinylpolypyrrolidone), ~110 μm particle size
Sigma-Aldrich
Povidone, meets USP testing specifications
USP
Crospovidone, United States Pharmacopeia (USP) Reference Standard
Povidone, European Pharmacopoeia (EP) Reference Standard
Crospovidone, European Pharmacopoeia (EP) Reference Standard
Supelco
Povidone, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Povidone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Polyvinylpyrrolidone, average Mw ~1,300,000 by LS
Sigma-Aldrich
Polyvinylpyrrolidone, K 30
Sigma-Aldrich
Polyvinylpyrrolidone, powder, average Mw ~55,000
Sigma-Aldrich
Polyvinylpyrrolidone, suitable for plant cell culture, average mol wt 10,000
Sigma-Aldrich
Polyvinylpyrrolidone, mol wt (number average molecular weight Mn 360kDa)
Sigma-Aldrich
Polyvinylpyrrolidone, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Polyvinylpyrrolidone, average mol wt 10,000
Sigma-Aldrich
Polyvinylpyrrolidone, Molecular Biology, nucleic acid hybridization tested, mol wt 360,000
Sigma-Aldrich
Polyvinylpyrrolidone, average mol wt 40,000
Sigma-Aldrich
Polyvinylpyrrolidone, K 25, tested according to Ph. Eur.
Sigma-Aldrich
Polyvinylpyrrolidone, powder, average Mw ~29,000
Sigma-Aldrich
Polyvinylpyrrolidone solution, K 60, 45% in H2O
Sigma-Aldrich
Polyvinylpyrrolidone, K 90
Sigma-Aldrich
Polyvinylpyrrolidone, Vetec, reagent grade, average mol wt 360,000