Merck
Zinc and myocardial ischemia/reperfusion injury.
Biometals : an international journal on the role of metal ions in biology, biochemistry, and medicine (2013-08-29)
Zhelong Xu, Juan Zhou
ABSTRACT
As an important trace element, zinc is required for the normal cellular structure and function, and impairment of zinc homeostasis is associated with a variety of health problems including cardiovascular disease. Zinc homeostasis is regulated through zinc transporters, zinc binding molecules, and zinc sensors. Zinc also plays a critical role in cellular signaling. Studies have documented that zinc homeostasis is impaired by ischemia/reperfusion in the heart and zinc dyshomeostasis may play a role in the pathogenesis of myocardial ischemia/reperfusion injury. Both exogenous and endogenously released zinc may play an important role in cardioprotection against ischemia/reperfusion injury. The goal of this review is to summarize the current understanding of the roles of zinc homeostasis and zinc signaling in myocardial ischemia/reperfusion injury.
Sigma-Aldrich
Zinc, dust, <10 μm, ≥98%
Sigma-Aldrich
Zinc, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.9%, granular
Sigma-Aldrich
Zinc preparation, 5 g/dL Zn+2 in THF, highly reactive Rieke®metal
Sigma-Aldrich
Zinc, pieces, 2-14 mesh, 99.9% trace metals basis
Sigma-Aldrich
Zinc, shot, <12 mm, 99.99% trace metals basis
Sigma-Aldrich
Zinc, foil, thickness 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Zinc, foil, thickness 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Zinc, shot, 5 mm, 99.999% trace metals basis
Sigma-Aldrich
Zinc, sticks, diam. 7-10 mm, 99.97% trace metals basis
Sigma-Aldrich
Zinc, granular, 20-30 mesh, ACS reagent, ≥99.8%
Sigma-Aldrich
Zinc, nanopowder, 40-60 nm avg. part. size, ≥99% trace metals basis
Zinc, foil, 15mm disks, thickness 0.05mm, as rolled, 99.99+%
Zinc, foil, 0.5m coil, thickness 0.20mm, as rolled, 99.95+%
Zinc, foil, 25mm disks, thickness 0.175mm, as rolled, 99.95+%
Zinc, foil, 15mm disks, thickness 0.0025mm, 99.9%
Zinc, foil, 10mm disks, thickness 0.025mm, as rolled, 99.95+%
Zinc, foil, 50x50mm, thickness 6mm, as rolled, 99.99+%
Zinc, foil, 25x25mm, thickness 3mm, as rolled, 99.99+%
Zinc, foil, 0.5m coil, thickness 0.075mm, as rolled, 99.95+%
Zinc, foil, 10mm disks, thickness 0.25mm, as rolled, 99.99+%
Zinc, foil, 25x25mm, thickness 6mm, as rolled, 99.99+%
Zinc, foil, 1m coil, thickness 0.025mm, as rolled, 99.95+%
Zinc, foil, 0.5m coil, thickness 0.125mm, as rolled, 99.95+%
Zinc, foil, 50x50mm, thickness 0.15mm, as rolled, 99.95+%
Zinc, foil, 0.5m coil, thickness 0.05mm, as rolled, 99.95+%
Zinc, foil, 15mm disks, thickness 0.1mm, as rolled, 99.95+%
Zinc, foil, 10mm disks, thickness 0.35mm, as rolled, 99.95+%
Zinc, foil, 0.5m coil, thickness 2.0mm, as rolled, 99.95+%
Zinc, foil, 0.3m coil, thickness 0.175mm, as rolled, 99.95+%
Zinc, foil, 15mm disks, thickness 0.5mm, as rolled, 99.95+%