Merck
Endoscopic ultrasound-guided ethanol ablation therapy for tumors.
World journal of gastroenterology (2013-06-27)
Wen-Ying Zhang, Zhao-Shen Li, Zhen-Dong Jin
ABSTRACT
Endoscopic ultrasonography (EUS) has evolved into a useful therapeutic tool for treating a broad range of tumors since being introduced into clinical practice as a diagnostic modality nearly three decades ago. In particular, EUS-guided fine-needle injection has proven a successful minimally invasive approach for treating benign lesions such as pancreatic cysts, relieving pancreatic pain through celiac plexus neurolysis, and controlling local tumor growth of unresectable malignancies by direct delivery of anti-tumor agents. One such ablative agent, ethanol, is capable of safely ablating solid or cystic lesions in hepatic tissues via percutaneous injection. Recent research and clinical interest has focused on the promise of EUS-guided ethanol ablation as a safe and effective method for treating pancreatic tumor patients with small lesions or who are poor operative candidates. Although it is not likely to replace radical resection of localized lesions or systemic treatment of metastatic tumors in all patients, EUS-guided ablation is an ideal method for patients who refuse or are not eligible for surgery. Moreover, this treatment modality may play an active role in the development of future pancreatic tumor treatments. This article reviews the most recent clinical applications of EUS-guided ethanol ablation in humans for treating pancreatic cystic tumors, pancreatic neuroendocrine tumors, and metastatic lesions.
Product Description
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.003% water
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Reagent Alcohol, reagent grade
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.005% water
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Reagent Alcohol, suitable for HPLC
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 2% 2-butanone, A15 MEK1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, ACS reagent, prima fine spirit, without additive, F15 o1
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, HPLC/spectrophotometric grade
Sigma-Aldrich
Ethanol, puriss. p.a., absolute, ≥99.8% (GC)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, for molecular biology