Merck
  • Targeting mitochondrial dysfunction and neurodegeneration by means of coenzyme Q10 and its analogues.
Targeting mitochondrial dysfunction and neurodegeneration by means of coenzyme Q10 and its analogues.
Current medicinal chemistry (2011-08-10)
D Orsucci, M Mancuso, E Caldarazzo Ienco, A LoGerfo, G Siciliano
ABSTRACT
Coenzyme Q10 is a small electron carrier of the respiratory chain with antioxidant properties, widely used for the treatment of mitochondrial disorders. Mitochondrial diseases are neuromuscular disorders caused by impairment of the respiratory chain and increased generation of reactive oxygen species. Coenzyme Q10 supplementation is fundamental in patients with primary coenzyme Q10 deficiency. Furthermore, coenzyme Q10 and its analogues, idebenone and mitoquinone (or MitoQ), have been also used in the treatment of other neurogenetic/neurodegenerative disorders. In Friedreich ataxia idebenone may reduce cardiac hypertrophy and, at higher doses, also improve neurological function. These compounds may also play a potential role in other conditions which have been linked to mitochondrial dysfunction, such as Parkinson disease, Huntington disease, amyotrophic lateral sclerosis and Alzheimer disease. This review introduces mitochondrial disorders and Friedreich ataxia as two paradigms of the tight links existing between oxidative stress, respiratory chain dysfunction and neurodegeneration, and focuses on current and emerging therapeutic uses of coenzyme Q10 and idebenone in neurology.
Sigma-Aldrich
Coenzyme Q10, ≥98% (HPLC)
USP
Ubidecarenone, United States Pharmacopeia (USP) Reference Standard
Supelco
Coenzyme Q10, analytical standard
Ubidecarenone for system suitability, European Pharmacopoeia (EP) Reference Standard
Ubidecarenone, European Pharmacopoeia (EP) Reference Standard
USP
Ubidecarenone for System Suitability, United States Pharmacopeia (USP) Reference Standard