Merck
Intrathecal baclofen in multiple sclerosis: too little, too late?
Multiple sclerosis (Houndmills, Basingstoke, England) (2011-02-02)
April Erwin, Mark Gudesblatt, Francois Bethoux, Susan E Bennett, Stephen Koelbel, Robert Plunkett, Saud Sadiq, Valerie L Stevenson, Ann-Marie Thomas, Carlo Tornatore, Mauro Zaffaroni, Mary Hughes
ABSTRACT
The majority of patients with multiple sclerosis (MS) have symptoms of spasticity that increasingly impair function as the disease progresses. With appropriate treatment, however, quality of life can be improved. Oral antispasticity medications are useful in managing mild spasticity but are frequently ineffective in controlling moderate to severe spasticity, because patients often cannot tolerate the adverse effects of increasing doses. Intrathecal baclofen (ITB) therapy can be an effective alternative to oral medications in patients who have a suboptimal response to oral medications or who cannot tolerate dose escalation or multidrug oral regimens. ITB therapy may be underutilized in the MS population because clinicians (a) are more focused on disease-modifying therapies rather than symptom control, (b) underestimate the impact of spasticity on quality of life, and (c) have concerns about the cost and safety of ITB therapy. Delivery of ITB therapy requires expertly trained staff and proper facilities for pump management. This article summarizes the findings and recommendations of an expert panel on the use of ITB therapy in the MS population and the role of the physician and comprehensive care team in patient selection, screening, and management.
Product Description
Baclofen, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
(±)-Baclofen, ≥98% (HPLC), solid