Merck
  • An overview of the visualisation and quantitation of low and high MW DNA adducts using the trapped in agarose DNA immunostaining (TARDIS) assay.
An overview of the visualisation and quantitation of low and high MW DNA adducts using the trapped in agarose DNA immunostaining (TARDIS) assay.
Mutagenesis (2010-11-12)
Ian G Cowell, Michael J Tilby, Caroline A Austin
ABSTRACT
The ability to detect and quantify specific DNA adducts benefits genome stability research, drug development and the evaluation of environmental mutagens. The trapped in agarose DNA immunostaining (TARDIS) assay was developed as a means of detecting and quantifying melphalan and cisplatin DNA adducts at the single-cell level and has since been adapted to quantify topoisomerase-DNA complexes. The method relies on salt-detergent extraction of agarose-embedded cells. Genomic DNA and any covalently attached molecules remain in place in the agarose, while other cellular constituents are removed. Drug-DNA or topoisomerase-DNA complexes are then detected and quantified by sensitive immunofluorescence using adduct-specific antibodies. Here, we give a perspective of the TARDIS assay including a comparison with other methods for quantifying topoisomerase-DNA covalent complexes and provide technical details required to set up and perform the assay.
Product Description
Sepharose 6B, Cytiva 17-0110-01, pack of 1 L
Sigma-Aldrich
Agarose, High EEO
Sigma-Aldrich
Agarose, High EEO, for molecular biology
Sigma-Aldrich
Agarose, BioReagent, for molecular biology, low EEO
Sigma-Aldrich
Agarose, Special High EEO
Sigma-Aldrich
Agarose, Ultra-low Gelling Temperature, molecular biology grade
Sigma-Aldrich
Agarose, For pulsed field electrophoresis running gel
Sigma-Aldrich
Agarose, BioReagent, for molecular biology, Wide range/Standard 3:1
Sigma-Aldrich
Agarose, Type I-A, low EEO
Sigma-Aldrich
Agarose, Type I, low EEO
Sigma-Aldrich
Sepharose 6B, 6% Beaded Agarose, 45-165 μm (wet), fractionation range 10,000-1,000,000 Da (dextrans)
Sigma-Aldrich
Agarose, Low EEO
Sigma-Aldrich
Agarose, for molecular biology
Sigma-Aldrich
Agarose, Ultra-low Gelling Temperature
Sigma-Aldrich
Agarose, High Gelling Temperature
Sigma-Aldrich
Agarose, Wide range, for molecular biology
Sigma-Aldrich
Agarose, Type IV, Special High EEO
Sigma-Aldrich
Agarose, Medium EEO, for molecular biology
Sigma-Aldrich
Agarose, Low EEO, for Immunoelectrophoresis
Sigma-Aldrich
Agarose, Type II-A, Medium EEO
Sepharose 4B, Cytiva 17-0120-01, pack of 1 L
Sigma-Aldrich
Sepharose 4B, 45-165 μm bead diameter