Merck
Allergic contact dermatitis caused by farnesol: clinical relevance.
Cutaneous and ocular toxicology (2010-09-23)
Sarah Gilpin, Howard Maibach
ABSTRACT
The fragrance material farnesol is cited as an infrequent but important cause of allergic contact dermatitis (ACD). It is included in the fragrance mix II patch series and requires labeling in the European Union if it is used in a consumer product. To review the existing literature to determine the causative role of farnesol in clinical contact allergy. Survey of the literature on farnesol studies; predictive and clinical elicitation tests in case reports, reviews, and abstracts. Predictive animal studies demonstrated in most cases that farnesol was a nonsensitizer. However, 2 local lymph node assays (LLNAs) indicated strong sensitization potential. Predictive human test data indicated a low potential, if any, for sensitization in human tests with farnesol at 10% or 12%. A few clinical reports indicated low-level allergy or questionable reactions to farnesol, with 5% being the most commonly used. There were also reports in which no reactions were seen. Predictive testing on farnesol in animals shows conflicting results depending on the study methodology used. Human predictive patch-test data also had gaps that prevented it from being definitive in pointing to a causative relationship between farnesol and contact dermatitis. The real sensitizing potential of a material can best be determined by evaluating the clinical and epidemiological data so as to help resolve the conflicting animal and human predictive test data. This literature and scoring exercise showed that predictive and clinical elicitation data do not document a clear causative determination that farnesol is a frequent contact allergen. Detailed clinical relevance and patient studies should clarify the clinical problem farnesol represents.
Sigma-Aldrich
Farnesol, mixture of isomers, ≥95%, stabilized, FG
Sigma-Aldrich
Farnesol, 95%
Supelco
Farnesol, analytical standard, stabilized, mixture of isomers