Merck
  • Experience of nitisinone for the pharmacological treatment of hereditary tyrosinaemia type 1.
Experience of nitisinone for the pharmacological treatment of hereditary tyrosinaemia type 1.
Expert opinion on pharmacotherapy (2008-04-22)
Saikat Santra, Ulrich Baumann
ABSTRACT
Hereditary tyrosinaemia type 1 is a rare inherited metabolic condition, which leads to a fatal multisystemic disease in childhood. Since 1992, nitisinone - a compound developed from work on triketone herbicides - has become an effective pharmacological treatment by inhibiting the enzyme 4-hydroxyphenylpyruvate dioxygenase. This review examines recent pharmacological and clinical literature on nitisinone, and assesses its impact as a pharmacological treatment for hereditary tyrosinaemia type 1. English language literature from MedLine and EmBase for nitisinone was searched from 1990 to 2008 for all papers relevant to the use of nitisinone in hereditary tyrosinaemia type 1. Nitisinone can prevent the development of liver disease and significantly reduce the risk of developing hepatocellular carcinoma; however, vigorous surveillance for the development of HCC needs to be continued lifelong.
Sigma-Aldrich
Nitisinone, ≥95% (HPLC)