Merck
Acetoin metabolism in bacteria.
Critical reviews in microbiology (2007-06-15)
Zijun Xiao, Ping Xu
ABSTRACT
Acetoin is an important physiological metabolite excreted by many microorganisms. The excretion of acetoin, which can be diagnosed by the Voges Proskauer test and serves as a microbial classification marker, has its vital physiological meanings to these microbes mainly including avoiding acification, participating in the regulation of NAD/NADH ratio, and storaging carbon. The well-known anabolism of acetoin involves alpha-acetolactat synthase and alpha-acetolactate decarboxylase; yet its catabolism still contains some differing views, although much attention has been focused on it and great advances have been achieved. Current findings in catabolite control protein A (CcpA) mediated carbon catabolite repression may provide a fuller understanding of the control mechanism in bacteria. In this review, we first examine the acetoin synthesis pathways and its physiological meanings and relevancies; then we discuss the relationship between the two conflicting acetoin cleavage pathways, the enzymes of the acetoin dehydrogenase enzyme system, major genes involved in acetoin degradation, and the CcpA mediated acetoin catabolite repression pathway; in the end we discuss the genetic engineering progresses concerning applications. To date, this is the first integrated review on acetoin metabolism in bacteria, especially with regard to catabolic aspects. The apperception of the generation and dissimilation of acetoin in bacteria will help provide a better understanding of microbial strategies in the struggle for resources, which will consequently better serve the utilization of these microbes.
Sigma-Aldrich
Acetoin, natural, ≥95%, FG
Supelco
Acetoin, analytical standard
Sigma-Aldrich
Acetoin, May exist as crystalline dimer
Sigma-Aldrich
Acetoin, primarily dimer, ≥95%, FG