Merck
  • Edge plane pyrolytic graphite electrodes in electroanalysis: an overview.
Edge plane pyrolytic graphite electrodes in electroanalysis: an overview.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry (2005-12-02)
Craig E Banks, Richard G Compton
ABSTRACT
The recent development, behavior and scope of edge plane pyrolytic graphite electrodes in electroanalysis are overviewed. Similarities to, and advantages, over multi-walled CNT modified electrodes are noted and the wide scope of applications, ranging through gas sensing, stripping voltammetry and biosensing, illustrated.
Sigma-Aldrich
Graphite, powder, <45 μm, ≥99.99% trace metals basis
Sigma-Aldrich
Graphite, rod, L 150 mm, diam. 6 mm, 99.995% trace metals basis
Sigma-Aldrich
Graphite, rod, L 150 mm, diam. 3 mm, low density, 99.995% trace metals basis
Sigma-Aldrich
Graphite, powder, <150 μm, 99.99% trace metals basis
Sigma-Aldrich
Graphite, powder, <20 μm, synthetic
Sigma-Aldrich
Graphite, flakes