Merck
  • Shifting paradigms in prostate cancer; docetaxel plus low-dose prednisone - finally an effective chemotherapy.
Shifting paradigms in prostate cancer; docetaxel plus low-dose prednisone - finally an effective chemotherapy.
European journal of cancer (Oxford, England : 1990) (2005-03-02)
Ronald de Wit
ABSTRACT
Until now, the use of systemic chemotherapy for advanced androgen-independent prostate cancer has had very little to offer to patients. However, in 2004, two large randomised trials investigating docetaxel vs. mitoxantrone have both demonstrated survival improvements, and, in one of the trails, improvements in important secondary clinical outcome measures such as pain relief and quality of life measurements. In this current perspective, these two trials are summarised and discussed. It is concluded that, docetaxel every 3 weeks plus low-dose prednisone can be considered standard treatment for patients with androgen-independent disease.
Sigma-Aldrich
Prednisone, ≥98%
Pricing and availability is not currently available.
Supelco
Prednisone, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
USP
Prednisone, United States Pharmacopeia (USP) Reference Standard
Pricing and availability is not currently available.