Merck
Get up to 22% off for Pi Day until 3/26.Save Now
  • Pyridine N-oxide derivatives: unusual anti-HIV compounds with multiple mechanisms of antiviral action.
Pyridine N-oxide derivatives: unusual anti-HIV compounds with multiple mechanisms of antiviral action.
The Journal of antimicrobial chemotherapy (2005-01-15)
Jan Balzarini, Miguel Stevens, Erik De Clercq, Dominique Schols, Christophe Pannecouque
ABSTRACT
Pyridine N-oxide derivatives represent a new class of anti-HIV compounds, for which some members exclusively act through inhibition of HIV-1 reverse transcriptase and thus characteristically behave as non-nucleoside reverse transcriptase inhibitors. Other members act, additionally or alternatively, at a post-integrational event in the replication cycle of HIV, that is, at the level of HIV gene expression. Repeated administration of one of the prototype compounds (JPL-32) to DBA/2 and hu-PBMC-SCID mice demonstrated, in the absence of any acute toxicity, protective activity against HIV-induced destruction of CD4 human T lymphocytes.
Sigma-Aldrich
Pyridine N-oxide, 95%