Merck
  • Ketorolac tromethamine: an oral/injectable nonsteroidal anti-inflammatory for postoperative pain control.
Ketorolac tromethamine: an oral/injectable nonsteroidal anti-inflammatory for postoperative pain control.
Journal of oral and maxillofacial surgery : official journal of the American Association of Oral and Maxillofacial Surgeons (1992-12-01)
R J Redden
ABSTRACT
Ketorolac tromethamine is a new injectable/oral nonsteroidal anti-inflammatory analgesic with no apparent opiate receptor activity that has been administered alone and in combination with other opiate analgesics for the treatment of postoperative pain. The drug has shown promise in analgesic comparisons with morphine sulfate; it lacks the effects of respiratory depression and nausea and vomiting usually associated with narcotic agents. Intramuscular ketorolac may be particularly useful with those patients who have respiratory disease and patients being dismissed following short ambulatory or private-office anesthetic procedures.
Sigma-Aldrich
Trizma® base, anhydrous, free-flowing, Redi-Dri, ≥99.9%
Trometamol, European Pharmacopoeia (EP) Reference Standard
Supelco
Tromethamine, pharmaceutical secondary standard, certified reference material
Ketorolac trometamol for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Trizma® base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Sigma-Aldrich
Tromethamine, meets USP testing specifications
Sigma-Aldrich
Trizma® base, ≥99.9% (titration), crystalline
Sigma-Aldrich
Trizma® base, BioXtra, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)
Supelco
Trizma® base, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Trizma® base, ≥99.0% (T)
Sigma-Aldrich
Trizma® base, BioUltra, for molecular biology, ≥99.8% (T)
Sigma-Aldrich
Trizma® base, puriss. p.a., ≥99.7% (T)
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ACS reagent, ≥99.8%
Supelco
Ketorolac Tromethamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ketorolac tris salt, ≥99%, crystalline
Sigma-Aldrich
Sigma 7-9®, ≥99% (titration), crystalline
SAFC
Tromethamine
Ketorolac trometamol, European Pharmacopoeia (EP) Reference Standard