Merck
  • Exuberant local tissue reaction to intramuscular injection of nandrolone decanoate (Deca-Durabolin)--a steroid compound in a sesame seed oil base--mimicking soft tissue malignant tumors: a case report and review of the literature.
Exuberant local tissue reaction to intramuscular injection of nandrolone decanoate (Deca-Durabolin)--a steroid compound in a sesame seed oil base--mimicking soft tissue malignant tumors: a case report and review of the literature.
Military medicine (1992-12-01)
N K Khankhanian, Y A Hammers, P Kowalski
ABSTRACT
We present an unusual pseudotumor that formed in reaction to self-administered intramuscular injections of an anabolic steroid, nandrolone decanoate (Deca-Durabolin) in a young soldier. The histopathologic features which closely mimicked several malignant neoplasms could have led to an incorrect diagnosis of malignancy and unnecessary extensive surgery. To our knowledge, this phenomenon has not been previously reported.
Sigma-Aldrich
19-Nortestosterone, ≥99.0% (HPLC)
Sigma-Aldrich
19-Nortestosterone
Nandrolone decanoate for peak identification, European Pharmacopoeia (EP) Reference Standard
Nandrolone decanoate, European Pharmacopoeia (EP) Reference Standard
Nandrolone decanoate for system suitability, European Pharmacopoeia (EP) Reference Standard