Merck
Biotechnological production of 2-phenylethanol.
Applied microbiology and biotechnology (2002-06-20)
M M W Etschmann, W Bluemke, D Sell, J Schrader
ABSTRACT
2-Phenylethanol (2-PE) is an important flavour and fragrance compound with a rose-like odour. Most of the world's annual production of several thousand tons is synthesised by chemical means but, due to increasing demand for natural flavours, alternative production methods are being sought. Harnessing the Ehrlich pathway of yeasts by bioconversion of L-phenylalanine to 2-PE could be an option, but in situ product removal is necessary due to product inhibition. This review describes the microbial production of 2-PE, and also summarizes the chemical syntheses and the market situation.
Product Description
Supelco
Phenylethyl Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Phenethyl alcohol, natural, ≥99%, FCC, FG
Sigma-Aldrich
2-Phenylethanol, ≥99.0% (GC)
Sigma-Aldrich
Phenethyl alcohol, ≥99%, FCC, FG