Merck
  • Potassium iodide and the wolff-chaikoff effect: relevance for the dermatologist.
Potassium iodide and the wolff-chaikoff effect: relevance for the dermatologist.
Journal of the American Academy of Dermatology (2000-02-25)
W R Heymann
ABSTRACT
The inhibition of organic binding of iodide in the thyroid gland by excess iodide, resulting in the cessation of thyroid hormone synthesis, is known as the Wolff-Chaikoff effect. This review explores the nature of the Wolff-Chaikoff effect, both in terms of its potential mechanisms and its relevance to dermatologists who use potassium iodide as a therapeutic agent.
Sigma-Aldrich
Potassium iodide, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Potassium iodide, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium iodide, suitable for plant cell culture
Sigma-Aldrich
Potassium iodide, AnhydroBeads, −10 mesh, 99.998% trace metals basis
Sigma-Aldrich
Potassium iodide, purum p.a., ≥99.0% (AT)
Sigma-Aldrich
Potassium iodide, tested according to Ph. Eur.
Sigma-Aldrich
Potassium iodide, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium iodide, puriss. p.a., reag. ISO, reag. Ph. Eur., ≥99.0%
Sigma-Aldrich
Potassium iodide, puriss., meets analytical specification of Ph. Eur. BP, USP, 99.0-100.5% (calc. to the dried substance)
Sigma-Aldrich
Potassium iodide, ReagentPlus®, 99%
Sigma-Aldrich
Potassium iodide, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Potassium iodide, ≥99.99% trace metals basis
Sigma-Aldrich
Gold etchant, standard