Merck
An unusual cause of breast lump: isolated tuberculosis of the breast.
The American journal of tropical medicine and hygiene (2014-05-09)
Partha Pal, Sisir Kumar Patra, Sayantan Ray
ABSTRACT
A 60-year-old woman presented with a painful lump of 4 months duration in her right breast. She denied any history of breast trauma or family history of breast cancer. Physical examination detected a firm, tender mass in the upper inner quadrant of her right breast without an associated sinus tract. Cytology from fine-needle aspiration of the swelling showed epithelioid cells, lymphohistocytic aggregates, and necrosis. Diagnosis was confirmed by the identification of acid-fast bacilli in tissue sections using Ziehl-Neelsen staining.
Product Description
Rifampicin, European Pharmacopoeia (EP) Reference Standard
Supelco
Isoniazid, analytical standard, ≥99% (TLC)
Supelco
Isoniazid, Vetec, reagent grade, 98%
Ethambutol for system suitability, European Pharmacopoeia (EP) Reference Standard
Isoniazid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ethambutol dihydrochloride, antimycobacterial
Sigma-Aldrich
Rifampicin, suitable for plant cell culture, BioReagent, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
Rifampicin, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
Pyrazinecarboxamide
Ethambutol hydrochloride, European Pharmacopoeia (EP) Reference Standard
Pyrazinamide, European Pharmacopoeia (EP) Reference Standard