Merck
  • Preparation of single phase Zn2TiO4 spinel from a new ZnTi layered double hydroxide precursor.
Preparation of single phase Zn2TiO4 spinel from a new ZnTi layered double hydroxide precursor.
Journal of nanoscience and nanotechnology (2014-04-18)
Jianye Song, Mingzhe Leng, Hongdi Xiao, Liangji Zhang, Yaowei Qin, Wanguo Hou, Na Du, Jianqiang Liu
ABSTRACT
A single-source ZnTi-layered double hydroxide precursor was used to prepare single phase Zn2TiO4. This approach involves two steps: the calcination of a ZnTi-layered double hydroxide precursor and selective leaching zinc oxide from the resultant calcined products. The materials were characterized by X-ray diffraction (XRD), scanning electron microscope (SEM) thermogravimetric and differential thermal analysis (TG-DTA), transmission electron microscope (TEM), surface area measurement and UVvis diffuse reflection spectroscopy. The results indicated that a single phase Zn2TiO4 could be successfully obtained from a ZnTi-layered double hydroxide precursor at a relatively low temperature in short calcination time. The TEM and SEM show that the diameter of Zn2TiO4 particles prepared at 900 degrees C is in the range of 20-100 nm and smaller than that prepared by the solid-state method. UV-Vis diffuse reflection spectroscopy demonstrates that the material has an energy bandgap around 3.7 eV.
Sigma-Aldrich
Titanium(IV) oxide, rutile, 99.995% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, ≥99.98% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, powder, <5 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, nanopowder, <100 nm particle size, 99.5% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, <001>, (single crystal substrate), ≥99.9% trace metals basis, L × W × thickness 10 mm × 10 mm × 0.5 mm
Sigma-Aldrich
Titanium(IV) oxide, mixture of rutile and anatase, nanopowder, <100 nm particle size (BET), 99.5% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, mixture of rutile and anatase, nanoparticles, <150 nm particle size (volume distribution, DLS), dispersion, 40 wt. % in H2O, 99.5% trace metals basis
Titanium, tube, 200mm, outside diameter 10.3mm, inside diameter 8.7mm, wall thickness 0.8mm, annealed, 99.6+%
Titanium, tube, 100mm, outside diameter 1.05mm, inside diameter 0.75mm, wall thickness 0.15mm, annealed, 99.6+%
Titanium, rod, 500mm, diameter 1.5mm, annealed, 99.6+%
Titanium, rod, 25mm, diameter 50mm, 99.99+%
Titanium, tube, 200mm, outside diameter 6.35mm, inside diameter 5.53mm, wall thickness 0.41mm, annealed, 99.6+%
Titanium, tube, 100mm, outside diameter 6.1mm, inside diameter 5.1mm, wall thickness 0.5mm, annealed, 99.6+%
Titanium, tube, 1000mm, outside diameter 4.25mm, inside diameter 3.75mm, wall thickness 0.25mm, annealed, 99.6+%
Titanium, tube, 1000mm, outside diameter 25.4mm, inside diameter 23.62mm, wall thickness 0.89mm, annealed, 99.6+%
Titanium, rod, 500mm, diameter 5mm, annealed, 99.6+%
Titanium, rod, 200mm, diameter 8.0mm, as drawn, 99.99+%
Titanium, rod, 200mm, diameter 5mm, annealed, 99.6+%
Titanium, tube, 200mm, outside diameter 2.03mm, inside diameter 1.55mm, wall thickness 0.24mm, annealed, 99.6+%
Titanium, tube, 200mm, outside diameter 1.6mm, inside diameter 1.2mm, wall thickness 0.2mm, hard, 99.6+%
Titanium, rod, 200mm, diameter 16mm, annealed, 99.6+%
Titanium, rod, 100mm, diameter 16mm, as drawn, 99.99+%
Titanium, rod, 100mm, diameter 10mm, annealed, 99.6+%
Titanium, microfoil, disks, 10mm, thinness 0.25μm, specific density 112.6μg/cm2, permanent mylar 3.5μm support, 99.6+%
Titanium, tube, 200mm, outside diameter 2.03mm, inside diameter 1.63mm, wall thickness 0.2mm, annealed, 99.6+%
Titanium, tube, 100mm, outside diameter 6.35mm, inside diameter 5.53mm, wall thickness 0.41mm, annealed, 99.6+%
Titanium, tube, 200mm, outside diameter 25.4mm, inside diameter 23.62mm, wall thickness 0.89mm, annealed, 99.6+%
Titanium, tube, 100mm, outside diameter 2.03mm, inside diameter 1.55mm, wall thickness 0.24mm, annealed, 99.6+%
Titanium, tube, 1000mm, outside diameter 1.6mm, inside diameter 1.2mm, wall thickness 0.2mm, hard, 99.6+%
Titanium, rod, 50mm, diameter 20mm, as drawn, 99.99+%