Merck
Get up to 22% off for Pi Day until 3/26.Save Now
Lanthanum(III) triflate catalyzed direct amidation of esters.
Organic letters (2014-03-26)
Hiroyuki Morimoto, Risa Fujiwara, Yuhei Shimizu, Kazuhiro Morisaki, Takashi Ohshima
ABSTRACT
Lanthanum trifluoromethanesulfonate is an effective single-component catalyst for synthesizing a variety of amides directly from esters and amines under mild conditions. Highly selective amidation of esters and amines, as well as catalyst-controlled amidation of esters, demonstrated the effectiveness of the catalyst system.
Sigma-Aldrich
Lanthanum, pieces
Sigma-Aldrich
Lanthanum, powder, −40 mesh, under oil, 99.9% trace rare earth metals basis
Lanthanum, foil, 25x25mm, thickness 0.1mm, as rolled, 99%
Sigma-Aldrich
Ethyl 6-chloropyridine-3-carboxylate, 97%