Merck
Pyridoxine responsiveness in novel mutations of the PNPO gene.
Neurology (2014-03-25)
Barbara Plecko, Karl Paul, Philippa Mills, Peter Clayton, Eduard Paschke, Oliver Maier, Oswald Hasselmann, Gudrun Schmiedel, Simone Kanz, Mary Connolly, Nicole Wolf, Eduard Struys, Sylvia Stockler, Lucia Abela, Doris Hofer
ABSTRACT
To determine whether patients with pyridoxine-responsive seizures but normal biomarkers for antiquitin deficiency and normal sequencing of the ALDH7A1 gene may have PNPO mutations. We sequenced the PNPO gene in 31 patients who fulfilled the above-mentioned criteria. We were able to identify 11 patients carrying 3 novel mutations of the PNPO gene. In 6 families, a homozygous missense mutation p.Arg225His in exon 7 was identified, while 1 family was compound heterozygous for a novel missense mutation p.Arg141Cys in exon 5 and a deletion c.279_290del in exon 3. Pathogenicity of the respective mutations was proven by absence in 100 control alleles and expression studies in CHO-K1 cell lines. The response to pyridoxine was prompt in 4, delayed in 2, on EEG only in 2, and initially absent in another 2 patients. Two unrelated patients homozygous for the p.Arg225His mutation experienced status epilepticus when switched to pyridoxal 5'-phosphate (PLP). This study challenges the paradigm of exclusive PLP responsiveness in patients with pyridoxal 5'-phosphate oxidase deficiency and underlines the importance of consecutive testing of pyridoxine and PLP in neonates with antiepileptic drug-resistant seizures. Patients with pyridoxine response but normal biomarkers for antiquitin deficiency should undergo PNPO mutation analysis.
Sigma-Aldrich
Pyridoxine, ≥98%
Supelco
Pyridoxine Hydrochloride (B6), analytical standard
Sigma-Aldrich
Pyridoxine hydrochloride, ≥98% (HPLC)
Sigma-Aldrich
Pyridoxine hydrochloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture
USP
Pyridoxine hydrochloride, United States Pharmacopeia (USP) Reference Standard
Pyridoxine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Pyridoxine hydrochloride, meets USP testing specifications
Supelco
Pyridoxine hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material