Merck
  • Periplasmic disulfide isomerase DsbC is involved in the reduction of copper binding protein CueP from Salmonella enterica serovar Typhimurium.
Periplasmic disulfide isomerase DsbC is involved in the reduction of copper binding protein CueP from Salmonella enterica serovar Typhimurium.
Biochemical and biophysical research communications (2014-03-25)
Bo-Young Yoon, Jin-Sik Kim, Si-Hyeon Um, Inseong Jo, Jin-Wook Yoo, Kangseok Lee, Yong-Hak Kim, Nam-Chul Ha
ABSTRACT
Salmonella enterica serovar Typhimurium (S. Typhimurium) is a facultative intracellular pathogen with the ability to survive and replicate in macrophages. Periplasmic copper binding protein CueP is known to confer copper resistance to S. Typhimurium, and has been implicated in ROS scavenge activity by transferring the copper ion to a periplasmic superoxide dismutase or by directly reducing the copper ion. Structural and biochemical studies on CueP showed that its copper binding site is surrounded by conserved cysteine residues. Here, we present evidence that periplasmic disulfide isomerase DsbC plays a key role in maintaining CueP protein in the reduced state. We observed purified DsbC protein efficiently reduced the oxidized form of CueP, and that it acted on two (Cys104 and Cys172) of the three conserved cysteine residues. Furthermore, we found that a surface-exposed conserved phenylalanine residue in CueP was important for this process, which suggests that DsbC specifically recognizes the residue of CueP. An experiment using an Escherichia coli system confirmed the critical role played by DsbC in the ROS scavenge activity of CueP. Taken together, we propose a molecular insight into how CueP collaborates with the periplasmic disulfide reduction system in the pathogenesis of the bacteria.
Product Description
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Iron, puriss. p.a., carbonyl-Iron powder, low in magnesium and manganese compounds, ≥99.5% (RT)
Sigma-Aldrich
Iron, powder, −325 mesh, 97%
Sigma-Aldrich
Iron, chips, 99.98% trace metals basis
Sigma-Aldrich
Iron, foil, thickness 0.1 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Protein Disulfide Isomerase from bovine liver, ≥100 units/mg protein, lyophilized powder
Iron, IRMM®, certified reference material, 0.5 mm wire
Iron, foil, 1m coil, thickness 0.5mm, hard, 99.5%
Iron, foil, 2m coil, thickness 0.125mm, hard, 99.5%
Iron, foil, 25mm disks, thickness 0.05mm, as rolled, 99.99+%
Iron, foil, 25mm disks, thickness 0.004mm, 99.85%
Iron, foil, 1m coil, thickness 0.05mm, hard, 99.5%
Iron, foil, 25mm disks, thickness 0.008mm, 99.85%
Iron, foil, 25mm disks, thickness 0.25mm, hard, 99.5%
Iron, foil, 10mm disks, thickness 0.9mm, as rolled, 99.5%
Iron, foil, 20m coil, thickness 0.075mm, hard, 99.5%
Iron, foil, 25mm disks, thickness 0.015mm, 99.85%
Iron, foil, 1m coil, thickness 0.20mm, hard, 99.5%
Iron, foil, 25mm disks, thickness 0.01mm, 99.99+%
Iron, foil, 4mm disks, thickness 0.009mm, 99.85%
Iron, foil, 2m coil, thickness 0.025mm, hard, 99.5%
Iron, foil, 20m coil, thickness 0.038mm, hard, 99.5%
Iron, foil, 1m coil, thickness 0.075mm, hard, 99.5%
Iron, foil, 15mm disks, thickness 0.05mm, as rolled, 99.99+%
Iron, foil, 10mm disks, thickness 1.0mm, as rolled, 99.5%
Iron, foil, 10mm disks, thickness 0.125mm, as rolled, 99.99+%
Iron, foil, 25mm disks, thickness 0.020mm, 99.85%
Iron, foil, 25mm disks, thickness 0.009mm, 99.85%
Iron, foil, 15mm disks, thickness 0.25mm, hard, 99.5%
Iron, foil, 4mm disks, thickness 0.004mm, 99.99+%