Merck
  • Fibroblast growth factor 21 and thyroid hormone show mutual regulatory dependency but have independent actions in vivo.
Fibroblast growth factor 21 and thyroid hormone show mutual regulatory dependency but have independent actions in vivo.
Endocrinology (2014-02-26)
Eleni M Domouzoglou, Ffolliott Martin Fisher, Inna Astapova, Elliott C Fox, Alexei Kharitonenkov, Jeffrey S Flier, Anthony N Hollenberg, Eleftheria Maratos-Flier
ABSTRACT
Thyroid hormone (TH) regulates fibroblast growth factor 21 (FGF21) levels in the liver and in the adipose tissue. In contrast, peripheral FGF21 administration leads to decreased circulating levels of TH. These data suggest that FGF21 and TH could interact to regulate metabolism. In the present study, we confirmed that TH regulates adipose and hepatic FGF21 expression and serum levels in mice. We next investigated the influence of TH administration on key serum metabolites, gene expression in the liver and brown adipose tissue, and energy expenditure in FGF21 knockout mice. Surprisingly, we did not observe any significant differences in the effects of TH on FGF21 knockout mice compared with those in wild-type animals, indicating that TH acts independently of FGF21 for the specific outcomes studied. Furthermore, exogenous FGF21 administration to hypothyroid mice led to similar changes in serum and liver lipid metabolites and gene expression in both hypothyroid and euthyroid mice. Thus, it appears that FGF21 and TH have similar actions to decrease serum and liver lipids despite having some divergent regulatory effects. Whereas TH leads to up-regulation in the liver and down-regulation in brown adipose tissue of genes involved in the lipid synthesis pathway (eg, fatty acid synthase (FASN) and SPOT14), FGF21 leads to the opposite changes in expression of these genes. In conclusion, TH and FGF21 act independently on the outcomes studied, despite their ability to regulate each other's circulating levels. Thus, TH and FGF21 may modulate the availability of each other in critical metabolic states.
Product Description
Thiamazole, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
2-Mercapto-1-methylimidazole, ≥99%
Supelco
Methimazole, VETRANAL®, analytical standard
Supelco
Methimazole, analytical standard
Sigma-Aldrich
L-Thyroxine, ≥98% (HPLC)
Sigma-Aldrich
L-Thyroxine, powder, BioReagent, suitable for cell culture
USP
Methimazole, United States Pharmacopeia (USP) Reference Standard
USP
Levothyroxine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Fibroblast Growth Factor from bovine pituitary, suitable for cell culture