Merck
  • The K(C) channel in the cbb3-type respiratory oxygen reductase from Rhodobacter capsulatus is required for both chemical and pumped protons.
The K(C) channel in the cbb3-type respiratory oxygen reductase from Rhodobacter capsulatus is required for both chemical and pumped protons.
Journal of bacteriology (2014-02-25)
Gülgez Gökçe Yıldız, Robert B Gennis, Fevzi Daldal, Mehmet Öztürk
ABSTRACT
The heme-copper superfamily of proton-pumping respiratory oxygen reductases are classified into three families (A, B, and C families) based on structural and phylogenetic analyses. Most studies have focused on the A family, which includes the eukaryotic mitochondrial cytochrome c oxidase as well as many bacterial homologues. Members of the C family, also called the cbb3-type oxygen reductases, are found only in prokaryotes and are of particular interest because of their presence in a number of human pathogens. All of the heme-copper oxygen reductases require proton-conducting channels to convey chemical protons to the active site for water formation and to convey pumped protons across the membrane. Previous work indicated that there is only one proton-conducting input channel (the K(C) channel) present in the cbb3-type oxygen reductases, which, if correct, must be utilized by both chemical protons and pumped protons. In this work, the effects of mutations in the K(C) channel of the cbb3-type oxygen reductase from Rhodobacter capsulatus were investigated by expressing the mutants in a strain lacking other respiratory oxygen reductases. Proton pumping was evaluated by using intact cells, and catalytic oxygen reductase activity was measured in isolated membranes. Two mutations, N346M and Y374F, severely reduced catalytic activity, presumably by blocking the chemical protons required at the active site. One mutation, T272A, resulted in a substantially lower proton-pumping stoichiometry but did not inhibit oxygen reductase activity. These are the first experimental data in support of the postulate that pumped protons are taken up from the bacterial cytoplasm through the K(C) channel.
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, viscosity 2,600-5,600 cP, 2 % in H2O(20 °C)(lit.)
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, viscosity 40-60 cP, 2 % in H2O(20 °C)(lit.)
USP
Hypromellose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, average Mn ~90,000
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, viscosity 80-120 cP, 2 % in H2O(20 °C)(lit.)
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, average Mn ~120,000
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, average Mn ~10,000
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, average Mn ~86,000
Sigma-Aldrich
Hypromellose, meets USP testing specifications
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose