Merck
  • The association between occurrence of plasmid-mediated quinolone resistance and ciprofloxacin resistance in Escherichia coli isolates of different origins.
The association between occurrence of plasmid-mediated quinolone resistance and ciprofloxacin resistance in Escherichia coli isolates of different origins.
Veterinary microbiology (2014-02-21)
Tong Yang, Zhenling Zeng, Lili Rao, Xiaojie Chen, Dandan He, Luchao Lv, Jing Wang, Li Zeng, Minsha Feng, Jian-Hua Liu
ABSTRACT
This study was performed to determine the prevalence of plasmid-mediated quinolone resistance (PMQR) determinants and characterize the ciprofloxacin resistance in Escherichia coli isolated from different sources in China. PMQR determinants were detected by PCR amplification and sequencing in 2297 E. coli isolates randomly collected from animals, food and humans during 2004 to 2011. MICs of ciprofloxacin were determined by agar dilution method. Of the 2297 E. coli isolates, 43.6% harbored at least one PMQR gene. The most common PMQR gene was oqxAB (29.3%), followed by qnr (13.6%), aac(6')-Ib-cr (11.6%), and qepA (3.3%). 12.0% isolates carried two or more PMQR genes. The prevalence of PMQR genes in food animal isolates increased over time, from 38.7% in 2004 to 69.8% in 2011. The prevalence of PMQR/ciprofloxacin resistance among isolates from pig, chicken, duck, companion animals, animal food and human volunteers were 65.2%/69.6%, 42.4%/60.0%, 59.4%/65.0%, 28.6%/57.5%, 29.3%/25.6%, and 14.0/8.7%, respectively. Most isolates carrying qnr along showed susceptible to ciprofloxacin, and only 21.6% the isolates exhibited resistance to ciprofloxacin, which was significantly lower than those carrying other PMQR genes (65.2-89.9%) and those that do not (43.1%) (p<0.01). In conclusion, high frequency of ciprofloxacin resistance and PMQR genes was observed among E. coli isolates of different origins in China, with oqxAB being the most frequent. qnr-positive E. coli isolates have relatively low ciprofloxacin resistance rate compared with other PMQR determinants-carrying isolates and PMQR-negative isolates.
Product Description
Supelco
Ciprofloxacin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ciprofloxacin, VETRANAL®, analytical standard
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)
Ciprofloxacin hydrochloride, European Pharmacopoeia (EP) Reference Standard
Ciprofloxacin, European Pharmacopoeia (EP) Reference Standard
Ciprofloxacin hydrochloride for peak identification, European Pharmacopoeia (EP) Reference Standard
USP
Ciprofloxacin hydrochloride, United States Pharmacopeia (USP) Reference Standard
Supelco
Ciprofloxacin HCl, Pharmaceutical Secondary Standard; Certified Reference Material