Merck
  • Bisphenol-A and diethylstilbestrol exposure induces the expression of breast cancer associated long noncoding RNA HOTAIR in vitro and in vivo.
Bisphenol-A and diethylstilbestrol exposure induces the expression of breast cancer associated long noncoding RNA HOTAIR in vitro and in vivo.
The Journal of steroid biochemistry and molecular biology (2014-02-19)
Arunoday Bhan, Imran Hussain, Khairul I Ansari, Samara A M Bobzean, Linda I Perrotti, Subhrangsu S Mandal
ABSTRACT
Antisense transcript, long non-coding RNA HOTAIR is a key player in gene silencing and breast cancer and is transcriptionally regulated by estradiol. Here, we have investigated if HOTAIR expression is misregulated by bisphenol-A (BPA) and diethylstilbestrol (DES). Our findings demonstrate BPA and DES induce HOTAIR expression in cultured human breast cancer cells (MCF7) as well as in vivo in the mammary glands of rat. Luciferase assay showed that HOTAIR promoter estrogen-response-elements (EREs) are induced by BPA and DES. Estrogen-receptors (ERs) and ER-coregulators such as MLL-histone methylases (MLL1 and MLL3) bind to the HOTAIR promoter EREs in the presence of BPA and DES, modify chromatin (histone methylation and acetylation) and lead to gene activation. Knockdown of ERs down-regulated the BPA and DES-induced expression of HOTAIR. In summary, our results demonstrate that BPA and DES exposure alters the epigenetic programming of the HOTAIR promoters leading to its endocrine disruption in vitro and in vivo.
Product Description
Supelco
Diethylstilbestrol, VETRANAL®, analytical standard
Sigma-Aldrich
Diethylstilbestrol, ≥99% (HPLC)
Diethylstilbestrol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Anti-MLL4 antibody produced in rabbit, IgG fraction of antiserum
Sigma-Aldrich
Bisphenol A, 97%
Supelco
Bisphenol A, ≥99%
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Supelco
Bisphenol A, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland