Merck
  • Effects of silver nanoparticles (NM-300K) on Lumbricus rubellus earthworms and particle characterization in relevant test matrices including soil.
Effects of silver nanoparticles (NM-300K) on Lumbricus rubellus earthworms and particle characterization in relevant test matrices including soil.
Environmental toxicology and chemistry (2013-12-10)
Merel J C van der Ploeg, Richard D Handy, Pauline L Waalewijn-Kool, Johannes H J van den Berg, Zahira E Herrera Rivera, Jan Bovenschen, Bastiaan Molleman, Johannes M Baveco, Peter Tromp, Ruud J B Peters, Gerwin F Koopmans, Ivonne M C M Rietjens, Nico W van den Brink
ABSTRACT
The impact of silver nanoparticles (AgNP; at 0 mg Ag/kg, 1.5 mg Ag/kg, 15.4 mg Ag/kg, and 154 mg Ag/kg soil) and silver nitrate (AgNO3 ; 15.4 mg Ag/kg soil) on earthworms, Lumbricus rubellus, was assessed. A 4-wk exposure to the highest AgNP treatment reduced growth and reproduction compared with the control. Silver nitrate (AgNO3 ) exposure also impaired reproduction, but not as much as the highest AgNP treatment. Long-term exposure to the highest AgNP treatment caused complete juvenile mortality. All AgNP treatments induced tissue pathology. Population modeling demonstrated reduced population growth rates for the AgNP and AgNO3 treatments, and no population growth at the highest AgNP treatment because of juvenile mortality. Analysis of AgNP treated soil samples revealed that single AgNP and AgNP clusters were present in the soil, and that the total Ag in soil porewater remained high throughout the long-term experiment. In addition, immune cells (coelomocytes) of earthworms showed sensitivity to both AgNP and AgNO3 in vitro. Overall, the present study indicates that AgNP exposure may affect earthworm populations and that the exposure may be prolonged because of the release of a dissolved Ag fraction to soil porewater.
Sigma-Aldrich
Silver, nanopowder, <100 nm particle size, contains PVP as dispersant, 99.5% trace metals basis
Sigma-Aldrich
Silver, evaporation slug, diam. × L 0.6 cm × 1.2 cm, 99.99% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 2.0 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 0.25 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, foil, thickness 0.025 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, foil, thickness 0.25 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 1.5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 0.5 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, powder, <45 μm, ≥99.99% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 0.25 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Silver, flakes, 10 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Silver, powder, 5-8 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Silver, powder, 2-3.5 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Silver nitrate on silica gel, extent of labeling: ~10 wt. % loading, +230 mesh
Sigma-Aldrich
Silver, granular, >250 μm, 99.99% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 2.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver nitrate solution, 2.5 % (w/v) AgNO3 in H2O
Silver, tube, 200mm, outside diameter 5.0mm, inside diameter 4.5mm, wall thickness 0.25mm, as drawn, 99.95+%
Silver, insulated wire, 5m, conductor diameter 0.25mm, insulation thickness 0.005mm, polyurethane insulation, 99.99%
Silver, insulated wire, 50m, conductor diameter 0.125mm, insulation thickness 0.0125mm, polyester insulation, 99.99%
Silver, insulated wire, 20m, conductor diameter 0.125mm, insulation thickness 0.015mm, PTFE (polytetrafluoroethylene) insulation, 99.99%
Silver, insulated wire, 5m, conductor diameter 0.25mm, insulation thickness 0.023mm, polyester insulation, 99.99%
Silver, insulated wire, 1m, conductor diameter 0.25mm, insulation thickness 0.005mm, polyurethane insulation, 99.99%
Silver, insulated wire, 50m, conductor diameter 0.125mm, insulation thickness 0.015mm, PTFE (polytetrafluoroethylene) insulation, 99.99%
Silver, insulated wire, 20m, conductor diameter 0.25mm, insulation thickness 0.015mm, epoxy insulation, 99.99%
Silver, insulated wire, 10m, conductor diameter 0.125mm, insulation thickness 0.0125mm, polyester insulation, 99.99%
Silver, insulated wire, 5m, conductor diameter 0.075mm, insulation thickness 0.010mm, PTFE (polytetrafluoroethylene) insulation, 99.99%
Silver, insulated wire, 20m, conductor diameter 0.025mm, insulation thickness 0.005mm, polyester insulation, 99.99%
Silver, insulated wire, 5m, conductor diameter 0.125mm, insulation thickness 0.0125mm, polyimide insulation, 99.9%
Silver, microfoil, disks, 25mm, thinness 0.1μm, specific density 110μg/cm2, permanent mylar 3.5μm support, 99.95+%