Merck
  • Corrosion behavior of tantalum-coated cobalt-chromium modular necks compared to titanium modular necks in a simulator test.
Corrosion behavior of tantalum-coated cobalt-chromium modular necks compared to titanium modular necks in a simulator test.
The Journal of arthroplasty (2013-10-09)
Ulrich Dorn, Daniel Neumann, Mario Frank
ABSTRACT
This study compared the corrosion behavior of tantalum-coated cobalt-chromium modular necks with that of titanium alloy modular necks at their junction to titanium-alloy femoral stem. Tests were performed in a dry assembly and two wet assemblies, one contaminated with calf serum and the other contaminated with calf serum and bone particles. Whereas the titanium modular neck tested in the dry assembly showed no signs of corrosion, the titanium modular necks tested in both wet assemblies showed marked depositions and corrosive attacks. By contrast, the tantalum-coated cobalt-chromium modular necks showed no traces of corrosion or chemical attack in any of the three assemblies. This study confirms the protective effect of tantalum coating the taper region of cobalt-chromium modular neck components, suggesting that the use of tantalum may reduce the risk of implant failure due to corrosion.
Product Description
Tantalum, foil, light tested, 50x50mm, thickness 0.01mm, 99.9%
Tantalum, foil, 100x100mm, thickness 0.100mm, annealed, 99.9%
Sigma-Aldrich
Tantalum, powder, 60-100 mesh, 99.9%
Sigma-Aldrich
Tantalum, foil, thickness 0.05 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Tantalum, powder, −325 mesh, 99.9% trace metals basis
Tantalum, foil, light tested, 100x100mm, thickness 0.015mm, 99.9%
Tantalum, foil, 50mm disks, thickness 0.0075mm, 99.9%
Tantalum, foil, light tested, 50x50mm, thickness 0.006mm, 99.9%
Tantalum, foil, 6mm disks, thickness 0.02mm, 99.9%
Tantalum, foil, 8mm disks, thickness 0.025mm, as rolled, 99.9%
Tantalum, foil, not light tested, 50x50mm, thickness 0.002mm, 99.9%
Tantalum, rod, 50mm, diameter 6.35mm, annealed, 99.9%
Tantalum, tube, 100mm, outside diameter 1.02mm, inside diameter 0.86mm, wall thickness 0.08mm, as drawn, 99.9%
Tantalum, foil, not light tested, 25x25mm, thickness 0.004mm, 99.9%
Tantalum, mesh, 40x40mm, nominal aperture 0.43mm, wire diameter 0.075mm, 50x50 wires/inch, open area 72%, plain weave mesh
Tantalum, foil, 8mm disks, thickness 0.006mm, 99.9%
Tantalum, foil, 6mm disks, thickness 0.0075mm, 99.9%
Tantalum, foil, 4mm disks, thickness 0.5mm, annealed, 99.9%
Tantalum, foil, light tested, 150x300mm, thickness 0.075mm, annealed, 99.9%
Tantalum, foil, 8mm disks, thickness 0.25mm, annealed, 99.99%
Tantalum, foil, 6mm disks, thickness 0.5mm, annealed, 99.9%
Tantalum, rod, 25mm, diameter 18mm, annealed, 99.9%
Tantalum, foil, light tested, 150x150mm, thickness 0.03mm, as rolled, 99.9%
Tantalum, rod, 100mm, diameter 1.7mm, annealed, 99.9%
Tantalum, tube, 1000mm, outside diameter 3.0mm, inside diameter 2mm, wall thickness 0.5mm, as drawn, 99.9%
Tantalum, foil, light tested, 25x25mm, thickness 0.003mm, 99.9%
Tantalum, rod, 500mm, diameter 2.0mm, annealed, 99.9%
Tantalum, rod, 200mm, diameter 1.5mm, annealed, 99.9%
Tantalum, foil, not light tested, 100x100mm, thickness 0.009mm, 99.9%
Tantalum, foil, light tested, 50x50mm, thickness 0.0075mm, 99.9%