Merck
  • Enzymatic trends of fructooligosaccharides production by microorganisms.
Enzymatic trends of fructooligosaccharides production by microorganisms.
Applied biochemistry and biotechnology (2013-12-18)
Mohd Anis Ganaie, Agbaje Lateef, Uma Shanker Gupta
ABSTRACT
Fructooligosaccharides are influential prebiotics that affect various physiological functions in such a way that they promote positive impact to health. They occur naturally in many fruits and vegetables in trace amounts. However, they are mainly produced commercially by the reaction of microbial enzymes with di- or polysaccharides, such as sucrose or inulin as a substrate. For maximum production of fructooligosaccharides on an industrial level, development of more enzymes with high activity and stability is required. This has attracted the attention of biotechnologists and microbiologists worldwide. This study aims to discuss the new trends in the production of fructooligosaccharide and its effect on numerous health qualities through which it creates great demand in the sugar market.
Sigma-Aldrich
Fructooligosaccharides from chicory, ≥90% (as FOS and inulin)
Pricing and availability is not currently available.