Merck
  • Synthesis of three diosgenyl saponins: dioscin, polyphyllin D, and balanitin 7.
Synthesis of three diosgenyl saponins: dioscin, polyphyllin D, and balanitin 7.
Carbohydrate research (1999-08-31)
S Deng, B Yu, Y Hui, H Yu, X Han
ABSTRACT
Dioscin, polyphyllin D, and balanitin 7, which belong to a group of structurally similar diosgenyl saponins with promising bioactivities, were synthesized by stepwise glycosylation.
Sigma-Aldrich
Hydrazine acetate, 97%