- Cryo-trapping/SPME/GC analysis of cheese aroma.
Cryo-trapping/SPME/GC analysis of cheese aroma.
Talanta (2008-10-31)
B Jaillais, V Bertrand, J Auger
PMID18967517
ABSTRACT
The advantages of combined cold-trapping of cheese volatiles and solid-phase microextraction-GC are demonstrated. This method is simple, cheap, compatible with classic identification equipment, and allows in-situ sampling.
Product Description
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm(PDMS, for use with autosampler, needle size 24 ga
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm(PDMS, needle size 24 ga, for use with manual holder
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), metal alloy fiber, df 100 μm(PDMS, needle size 23 ga, for use with autosampler
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm(PDMS, for use with manual holder, needle size 23 ga
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm(PDMS, for use with autosampler, needle size 23 ga