Merck
  • Solid-phase microextraction of hop volatiles. Potential use for determination and verification of hop varieties.
Solid-phase microextraction of hop volatiles. Potential use for determination and verification of hop varieties.
Journal of chromatography. A (2001-06-14)
M Kovacevic, M Kac
ABSTRACT
The composition of hop essential oil is an important tool for evaluation of hop quality. As each hop variety has a typical essential oil pattern (fingerprint), hop oil analyses can be used to distinguish between hop varieties. The headspace solid-phase microextraction (SPME) method as described in this contribution is a simple sample preparation technique and represents an alternative procedure for essential oil fingerprint determination. Different SPME parameters (extraction temperature, extraction time and sample mass) were studied and the results were compared with those obtained by the routine distillation method. It is shown that SPME results can be used for determination and verification of varieties grown in Slovenia by means of principal components analysis.
Product Description
Supelco
SPME Fiber Assembly, 100 μm PDMS, Metal Alloy (1 cm), needle size 23 ga, Autosampler, pk of 1, red hub
Supelco
SPME Fiber Assembly, 100 μm PDMS, Fused Silica (1 cm), needle size 23 ga, Manual Holder, pk of 3, red hub
Supelco
SPME Fiber Assembly, 100 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, red hub
Supelco
SPME Fiber Assembly, 100 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, red hub
Supelco
SPME Fiber Assembly, 100 μm PDMS, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, red hub
Supelco
SPME Portable Field Sampler, coating PDMS