Merck
  • The production of the eosinophil proteins ECP and EPX/EDN are regulated in a reciprocal manner.
The production of the eosinophil proteins ECP and EPX/EDN are regulated in a reciprocal manner.
APMIS : acta pathologica, microbiologica, et immunologica Scandinavica (2014-04-17)
Ulla-Britt Jönsson, Kristin Blom, Gunnemar Stålenheim, Lena Douhan Håkansson, Per Venge
ABSTRACT
Previous studies showed that the biological activity and the eosinophil content of eosinophil cationic protein (ECP, RNase 3) are determined by single-nucleotide polymorphisms (SNPs) in the ECP (RNase3) gene. In this study, we report the prevalence of a common SNP in the eosinophil protein x/eosinophil-derived neurotoxin (EPX/EDN, RNase2) and the association with the cellular contents of EPX/EDN and ECP. The genes were sequenced and the EPX/EDN405(G>C) rs2013109 SNPs were also determined by TaqMan 5′nuclease allelic discrimination assay. ECP and EPX/EDN in purified eosinophils or in whole blood extracts were analysed by sensitive immunoassays. The study included 379 non-allergic and allergic subjects. The genotype prevalence of the EPX/EDN405(G>C) polymorphism was GG 59%, GC 36% and CC 6%. The cellular contents of ECP and EPX/EDN were related in a reciprocal fashion with the sums of the protein contents being constant. The contents were associated with the ECP562(G>C) rs2233860 and EPX/EDN405(G>C) gene polymorphisms. The cellular content of eosinophil peroxidase (EPO) was not associated with the ECP and EPX/EDN genotypes. The prevalence of the EPX/EDN405(G>C) genotypes and the contents of the proteins were similar in non-allergic and allergic subjects.The production and storage of the two ancestral proteins, ECP and EPX/EDN likely share common regulatory mechanisms, which result in opposing productions of the two proteins.
Sigma-Aldrich
Ribonuclease A from bovine pancreas, Type I-AS, 50-100 Kunitz units/mg protein
Sigma-Aldrich
Ribonuclease A from bovine pancreas, Type III-A, ≥85% RNase A basis (SDS-PAGE), 85-140 Kunitz units/mg protein
Sigma-Aldrich
Ribonuclease A from bovine pancreas, Type II-A, ≥60% (SDS-PAGE), >= 60 Kunitz units/mg protein
Sigma-Aldrich
Ribonuclease A from bovine pancreas, Type I-A, powder, ≥60% RNase A basis (SDS-PAGE), ≥50 Kunitz units/mg protein
Sigma-Aldrich
Ribonuclease A from bovine pancreas, Type X-A, ≥90% (SDS-PAGE), ≥70 Kunitz units/mg protein
Sigma-Aldrich
Ribonuclease A from bovine pancreas, (Solution of 50% glycerol, 10mM Tris-HCL pH 8.0)
Sigma-Aldrich
Ribonuclease A from bovine pancreas, Type XII-A, ≥90% (SDS-PAGE), 75-125 Kunitz units/mg protein
Sigma-Aldrich
Ribonuclease A from bovine pancreas, for molecular biology, ≥70 Kunitz units/mg protein, lyophilized
Sigma-Aldrich
Ribonuclease B from bovine pancreas, BioReagent, ≥50 Kunitz units/mg protein, ≥80% (SDS-PAGE)
Sigma-Aldrich
Ribonuclease A from bovine pancreas, 90%, powder, white, ~80 U/mg
Sigma-Aldrich
Ribonuclease A from bovine pancreas, 4×cryst., ~70 U/mg (acc. to Kunitz)