Merck
  • Novel layer-by-layer structured nanofibrous mats coated by protein films for dermal regeneration.
Novel layer-by-layer structured nanofibrous mats coated by protein films for dermal regeneration.
Journal of biomedical nanotechnology (2014-04-17)
Shangjing Xin, Xueyong Li, Qun Wang, Rong Huang, Xiaoli Xu, Zhanjun Lei, Hongbing Deng
ABSTRACT
Layer-by-layer coating technique is effective in modifying the surface of nanofibrous mats, but overmuch film-coating makes the mats less porous to hardly suit the condition for tissue engineering. We developed novel nanofibrous mats layer-by-layer coated by silk fibroin and lysozyme on the cellulose electrospun template via electrostatic interaction. The film-coating assembled on the mats was not excessive because the charge of the proteins varied in the coating process due to different pH value. In addition, pure nature materials made the mats nontoxic, biodegradable and low-cost. The morphology and composition variation during layer-by-layer coating process was investigated and the results showed that the structure and thickness of film-coatings could be well-controlled. The antibacterial assay and in vitro cell experiments indicated that the mats could actively inhibit bacteria and exhibit excellent biocompatibility. In vivo implant assay further verified the mats cultured with human epidermal cells could promote wound healing and avoid wound infection. Therefore, these mats showed promising prospects when performed for dermal reconstruction.
Sigma-Aldrich
Lysozyme from human neutrophils, ≥95% (SDS-PAGE), lyophilized powder, ≥100,000 units/mg protein (E1%/280)
Sigma-Aldrich
Lysozyme human, Lysobac, recombinant, expressed in rice, ≥100,000 units/mg protein, lyophilized powder
Sigma-Aldrich
Lysozyme from chicken egg white, dialyzed, lyophilized, powder, ~100000 U/mg
Sigma-Aldrich
Lysozyme from chicken egg white, powder (crystalline), 70000-140000 U/mg
Sigma-Aldrich
Lysozyme from chicken egg white, 10 mg/mL
Sigma-Aldrich
Lysozyme from chicken egg white, aseptically filled
Sigma-Aldrich
Lysozyme chloride form from chicken egg white, Grade VI, ≥35,000 units/mg protein (E1%/282)
Sigma-Aldrich
Lysozyme from chicken egg white, For use as a marker in SDS-PAGE
Sigma-Aldrich
Lysozyme from chicken egg white, powder or granules, ≥39,000 units/mg protein
Sigma-Aldrich
Lysozyme from chicken egg white, BioUltra, lyophilized powder, ≥98% (SDS-PAGE), ≥40,000 units/mg protein