Merck
  • Glutathione-mediated reversibility of covalent modification of ubiquitin carboxyl-terminal hydrolase L1 by 1,2-naphthoquinone through Cys152, but not Lys4.
Glutathione-mediated reversibility of covalent modification of ubiquitin carboxyl-terminal hydrolase L1 by 1,2-naphthoquinone through Cys152, but not Lys4.
Chemico-biological interactions (2014-03-04)
Takashi Toyama, Yasuhiro Shinkai, Aki Yazawa, Hidenao Kakehashi, Toshiyuki Kaji, Yoshito Kumagai
ABSTRACT
Covalent modification of cellular proteins by electrophiles affects electrophilic signal transduction and the dysfunction of enzymes that is involved in cytotoxicity. We have recently found a unique reaction which restores glyceraldehyde-3-phosphate dehydrogenase (GAPDH) that has been modified by 1,2-naphthoquinone (1,2-NQ) through a glutathione (GSH)-dependent S-transarylation reaction. We report here that ubiquitin carboxyl-terminal hydrolase L1 (UCH-L1) undergoes the same reaction. Exposure of human neuroblastoma SH-SY5Y cells to 1,2-NQ after pretreatment with buthionine sulfoximine (BSO) to deplete GSH resulted in an enhancement of covalent modification of UCH-L1 by 1,2-NQ. With recombinant human UCH-L1, we demonstrated that UCH-L1 underwent arylation by 1,2-NQ through Cys152 and Lys4, thereby decreasing its catalytic activity. Addition of GSH to an incubation mixture of 1,2-NQ-UCH-L1 adduct partially restored this decline in enzyme activity which was accompanied by decreased covalent attachment of 1,2-NQ, together with production of 1,2-NQ-GSH adduct. UCH-L1 in which Lys4 was mutated exhibited a lower level of covalent modification and enzyme inhibition, but completely recovered after addition of GSH. Taken together, these results suggest that Cys152 modification in UCH-L1 by 1,2-NQ is reversible via GSH-mediated S-transarylation reaction whereas Lys4 modification by 1,2-NQ is irreversible by GSH. Because UCH-L1 dysfunction has been associated with neurodegeneration, the electrophilic modification of Lys rather than Cys in UCH-L1 may be implicated in such neurodegenerative diseases.
Product Description
Supelco
L-Lysine monohydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Cysteine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Lysine monohydrochloride, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Lysine Acetate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Cysteine, 97%
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
L-Cysteine, BioUltra, ≥98.5% (RT)
Sigma-Aldrich
L-Cysteine, ≥97%, FG
SAFC
L-Cysteine
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
L-Cysteine, from non-animal source, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
L-Cysteine, produced by Wacker Chemie AG, Burghausen, Germany, ≥98.0%
Sigma-Aldrich
L-Cysteine, Vetec, reagent grade, 97%
Sigma-Aldrich
L-Glutathione reduced, Vetec, reagent grade, ≥98%
Supelco
L-Lysine, analytical standard
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Lysine acetate salt, ≥98% (HPLC)
Sigma-Aldrich
L-Lysine, ≥98% (TLC)
Sigma-Aldrich
L-Lysine, crystallized, ≥98.0% (NT)
Lysine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Lysine acetate, European Pharmacopoeia (EP) Reference Standard
Glutathione, European Pharmacopoeia (EP) Reference Standard