Merck
Tissue expression pattern and polymorphism of G0S2 gene in porcine.
Gene (2014-02-04)
Yanzhi Jiang, Wangmin Cen, Shuhua Xing, Jianning Chen, Huaming Xu, Anxiang Wen, Li Zhu, Guoqing Tang, Mingzhou Li, Anan Jiang, Xuewei Li
ABSTRACT
Adipose triglyceride lipase (ATGL), catalyzing the initial step of hydrolysis of triacylglycerol (TAG) in adipocytes, has been known to be inhibited by G0/G1 switch protein 2 (G0S2). In this study, we determined tissue expression pattern and polymorphism of G0S2 gene in porcine. The results showed that the G0S2 transcript levels were very high in the liver and, to a lesser degree, in adipose tissues of greater omentum and suet fat; and low G0S2 transcript levels were observed in other tissues. A comparative study on the transcript levels between ATGL and G0S2 genes showed that ATGL transcript levels were high in all six adipose tissues, but negligible in the liver. Higher transcript levels were obtained for sows in adipose tissues of the inner layer of subcutaneous fat and suet fat, but higher expression values were found for boars in the liver, spleen, and stomach. 19 single nucleotide polymorphisms (SNPs), including 4 nonsynonymous SNPs (g.-307A>T, g.-394C>G, g.-565G>A, and g.-566T>C), were found in porcine G0S2 genomic DNA. Association analyses showed that the g.-565G>A and g.-742T>A SNPs were associated with back fat thickness (BFT). In conclusion, G0S2 mRNAs are abundantly expressed in porcine liver and adipose tissues of greater omentum and suet fat, and sex affects porcine G0S2 tissue transcript levels; meanwhile, the genetic diversity of porcine G0S2 gene is abundant and 2 SNPs are a genetic factor affecting BFT.
Sigma-Aldrich
Lipase from Rhizopus oryzae, powder (fine), ~10 U/mg
Sigma-Aldrich
Lipase from Candida rugosa, lyophilized, powder (fine), 15-25 U/mg
Sigma-Aldrich
Lipase from Aspergillus oryzae, lyophilized, powder, white, ~50 U/mg
Sigma-Aldrich
Lipase from Mucor miehei, powder, slightly brown, ~1 U/mg
Sigma-Aldrich
Lipase from Rhizopus niveus, powder (fine), ≥1.5 U/mg
Sigma-Aldrich
Lipase from Pseudomonas sp., Type XIII, lyophilized powder, ≥15 units/mg solid
Sigma-Aldrich
Lipase from Candida rugosa, Type VII, ≥700 unit/mg solid
Sigma-Aldrich
Lipase from Candida rugosa, lyophilized powder, ≥40,000 units/mg protein
Sigma-Aldrich
Lipase from Mucor miehei, lyophilized powder, ≥4,000 units/mg solid (using olive oil)
Sigma-Aldrich
Lipase from porcine pancreas, Type VI-S, ≥20,000 units/mg protein, lyophilized powder
Sigma-Aldrich
Lipase from wheat germ, Type I, lyophilized powder, 5-15 units/mg solid
Sigma-Aldrich
Lipase from Aspergillus oryzae, ≥20,000 U/g
Sigma-Aldrich
Lipase from Aspergillus oryzae, solution, ≥100,000 U/g, white, beige
Sigma-Aldrich
Lipase acrylic resin, ≥5,000 U/g, recombinant, expressed in Aspergillus niger
Sigma-Aldrich
Lipase from Candida rugosa, powder, yellow-brown, ≥2 U/mg
Sigma-Aldrich
Lipase from Candida sp., recombinant, expressed in Aspergillus niger
Sigma-Aldrich
Lipase A Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~2 U/mg
Sigma-Aldrich
Lipase B Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~9 U/mg
Sigma-Aldrich
Lipase from Aspergillus niger, powder (fine), ~200 U/g
Sigma-Aldrich
Lipase immobilized from Candida antarctica, beads, slightly brown, >2 U/mg
Sigma-Aldrich
Lipase from Mucor javanicus, lyophilized powder, ≥300 units/mg solid (using olive oil)
Sigma-Aldrich
Lipase from Pseudomonas cepacia, powder, light beige, ≥30 U/mg