Merck
  • The effect of PNPLA3 on fibrosis progression and development of hepatocellular carcinoma: a meta-analysis.
The effect of PNPLA3 on fibrosis progression and development of hepatocellular carcinoma: a meta-analysis.
The American journal of gastroenterology (2014-01-22)
Amit G Singal, Hema Manjunath, Adam C Yopp, Muhammad S Beg, Jorge A Marrero, Purva Gopal, Akbar K Waljee
ABSTRACT
The PNPLA3 rs738409 single-nucleotide polymorphism is known to promote nonalcoholic steatohepatitis (NASH), but its association with fibrosis severity and hepatocellular carcinoma (HCC) risk is less well-defined. The objectives of this study were to determine the association between PNPLA3 and liver fibrosis severity, HCC risk, and HCC prognosis among patients with liver disease. We performed a systematic literature review using the Medline, PubMed, Scopus, and Embase databases through May 2013 and a manual search of national meeting abstracts from 2010 to 2012. Two investigators independently extracted data on patient populations, study methods, and results using standardized forms. Pooled odds ratios (ORs), according to PNPLA3 genotype, were calculated using the DerSimonian and Laird method for a random effects model. Among 24 studies, with 9,915 patients, PNPLA3 was associated with fibrosis severity (OR 1.32, 95% confidence interval (CI) 1.20-1.45), with a consistent increased risk across liver disease etiologies. Among nine studies, with 2,937 patients, PNPLA3 was associated with increased risk of HCC in patients with cirrhosis (OR 1.40, 95% CI 1.12-1.75). On subgroup analysis, increased risk of HCC was demonstrated in patients with NASH or alcohol-related cirrhosis (OR 1.67, 95% CI 1.27-2.21) but not in those with other etiologies of cirrhosis (OR 1.33, 95% CI 0.96-1.82). Three studies, with 463 patients, do not support an association between PNPLA3 and HCC prognosis but are limited by heterogeneous outcome measures. For all outcomes, most studies were conducted in homogenous Caucasian populations, and studies among racially diverse cohorts are needed. PNPLA3 is associated with an increased risk of advanced fibrosis among patients with a variety of liver diseases and is an independent risk factor for HCC among patients with nonalcoholic steatohepatitis or alcohol-related cirrhosis.
Sigma-Aldrich
Lipase from Rhizopus oryzae, powder (fine), ~10 U/mg
Sigma-Aldrich
Lipase from Candida rugosa, lyophilized, powder (fine), 15-25 U/mg
Sigma-Aldrich
Lipase from Mucor miehei, powder, slightly brown, ~1 U/mg
Sigma-Aldrich
Lipase from Aspergillus oryzae, lyophilized, powder, white, ~50 U/mg
Sigma-Aldrich
Lipase from Candida rugosa, powder, yellow-brown, ≥2 U/mg
Sigma-Aldrich
Lipase from Rhizopus niveus, powder (fine), ≥1.5 U/mg
Sigma-Aldrich
Lipase from Mucor miehei, lyophilized powder, ≥4,000 units/mg solid (using olive oil)
Sigma-Aldrich
Lipase from Candida rugosa, Type VII, ≥700 unit/mg solid
Sigma-Aldrich
Lipase from Pseudomonas sp., Type XIII, lyophilized powder, ≥15 units/mg solid
Sigma-Aldrich
Lipase from porcine pancreas, Type VI-S, ≥20,000 units/mg protein, lyophilized powder
Sigma-Aldrich
Lipase from wheat germ, Type I, lyophilized powder, 5-15 units/mg solid
Sigma-Aldrich
Lipase from Candida rugosa, lyophilized powder, ≥40,000 units/mg protein
Sigma-Aldrich
Lipase from Aspergillus oryzae, solution, ≥100,000 U/g, white, beige
Sigma-Aldrich
Lipase from Aspergillus oryzae, ≥20,000 U/g
Sigma-Aldrich
Lipase acrylic resin, ≥5,000 U/g, recombinant, expressed in Aspergillus niger
Sigma-Aldrich
Lipase from Candida sp., recombinant, expressed in Aspergillus niger
Sigma-Aldrich
Lipase from Mucor javanicus, lyophilized powder, ≥300 units/mg solid (using olive oil)
Sigma-Aldrich
Lipase from Pseudomonas cepacia, powder, light beige, ≥30 U/mg
Sigma-Aldrich
Lipase from Aspergillus niger, powder (fine), ~200 U/g
Sigma-Aldrich
Lipase A Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~2 U/mg
Sigma-Aldrich
Lipase B Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~9 U/mg
Sigma-Aldrich
Lipase immobilized from Candida antarctica, beads, slightly brown, >2 U/mg